Skip to Content
Merck
All Photos(1)

Key Documents

PHR1133

Supelco

Cetyl Alcohol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-Hexadecanol, Cetyl alcohol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C0990000
traceable to USP 1103003

vapor density

8.34 (vs air)

vapor pressure

<0.01 mmHg ( 43 °C)

API family

cetyl alcohol

CofA

current certificate can be downloaded

autoignition temp.

483 °F

expl. lim.

8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

179-181 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Cetyl Alcohol may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7153 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The application of chromatography to the analysis of pharmaceutical creams.
Van de Vaart FJ, et al.
Chromatographia, 16(1), 247-250 (1982)
HPLC-fluorescence determination of chlorocresol and chloroxylenol in pharmaceuticals.
Gatti R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 16(3), 405-412 (1997)
Kerstin Wagner et al.
Chemistry and physics of lipids, 145(2), 119-127 (2006-12-26)
The monolayer structure of pure dipalmitoylphosphatidylcholine (DPPC) and equimolar mixtures of DPPC/n-hexadecanol (C(16)OH) and DPPC/dipalmitoylglycerol (DPG) are studied by the film balance technique and grazing incidence X-ray diffraction measurements. At 20 degrees C, the binary systems exhibit complete miscibility. In
Luca Lo Piccolo et al.
Applied and environmental microbiology, 77(4), 1204-1213 (2010-12-25)
Enzymes involved in oxidation of long-chain n-alkanes are still not well known, especially those in gram-positive bacteria. This work describes the alkane degradation system of the n-alkane degrader actinobacterium Gordonia sp. strain SoCg, which is able to grow on n-alkanes
Flávia C Carvalho et al.
Journal of pharmaceutical sciences, 99(5), 2367-2374 (2009-12-08)
The development of a controlled-release dosage form of zidovudine (AZT) is of crucial importance, in view of the pharmacokinetics of its toxic activity. A suitable drug delivery system could increase AZT bioavailability, reducing its dose-dependent side effects. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service