Skip to Content
Merck
All Photos(1)

Key Documents

PHR1076

Supelco

Metoprolol Tartrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(±)-Metoprolol (+)-tartrate salt, (±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 540
traceable to Ph. Eur. M1830000
traceable to USP 1441301

API family

metoprolol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2

InChI

1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

YGULWPYYGQCFMP-CEAXSRTFSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

Metoprolol Tartrate is a β-blocking drug, showing antihypertensive and anti-ischemic effects. It is widely used in the treatment of hypertension and helps in controlling ventricular response in patients with atrial fibrillation.

Application

Metoprolol Tartrate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations using densitometric technique and spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6905 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densitometric determination of metoprolol tartrate in pharmaceutical dosage forms
Vujic Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 15(5), 581-585 (1997)
A validated HPLC method for estimation of metoprolol in human plasma
Aqil M, et al.
Acta Chromatographica , 19, 130-130 (2007)
Current Cardiovascular Drugs, 15(4), 581-585 (2005)
AAS and spectrophotometric determination of propranolol HCl and metoprolol tartrate
El-Ries MA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(2), 179-187 (2000)
Thomas Kahan et al.
European journal of clinical investigation, 43(2), 141-151 (2013-01-03)
Although stable angina pectoris often carries a favourable prognosis, it remains important to identify patients with an increased risk of cardiovascular (CV) complications. Many new markers of disease activity and prognosis have been described. We evaluated whether common and easily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service