Skip to Content
Merck
All Photos(5)

Key Documents

P9375

Sigma-Aldrich

1,10-Phenanthroline monohydrate

reagent grade

Synonym(s):

o-Phenanthroline monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · H2O
CAS Number:
Molecular Weight:
198.22
Beilstein:
4008096
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

grade

reagent grade

form

powder

mp

100-104 °C (lit.)

solubility

methanol: 200 mM (Stock solution is stable for months at –20° C.)

SMILES string

O.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Looking for similar products? Visit Product Comparison Guide

General description

Phenanthroline is a heterocyclic organic compound, which forms complexes with metal ions. These complexes have wide applications in biochemistry, catalysis and analytical chemistry. It complexes with iron and is exploited as a redox indicator.

Application

1,10-Phenanthroline monohydrate has been used:
  • as an inhibitor of metalloproteinase (MMP) in MDA-MB-231 breast cancer cells
  • as a negative control in in situ zymography of mice heart samples
  • in in vitro resveratrol chelation and reducing activities assays to chelate iron

When complexed with copper, it possesses nuclease activity that has been used to study DNA-protein interactions.

Biochem/physiol Actions

Metalloprotease inhibitor; chelates iron, zinc and other divalent metals. Effective concentration 1-10 mM.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resveratrol attenuates iron-induced toxicity in a chronic post-treatment paradigm in Caenorhabditis elegans
Soares MV, et al.
Free Radical Research, 52(9), 939-951 (2018)
Katy Satué et al.
Acta veterinaria Hungarica, 68(1), 79-84 (2020-05-10)
In women and females of different species of laboratory animals, oestrogens stimulate the renin-angiotensin-aldosterone system (RAAS) by increasing tissue and circulating levels of angiotensinogen and renin during the preovulatory period. Progesterone and cortisol compete with aldosterone for mineralocorticoid receptors, which
Central Role of Phenanthroline Mono-N-oxide in the Decomposition Reactions of Tris (1, 10-phenanthroline) iron (II) and-iron (III) Complexes
Beller G, et al.
Inorganic Chemistry, 49(9), 3968-3970 (2010)
The transmodulation of HER2 and EGFR by substance P in breast cancer cells requires c-Src and metalloproteinase activation
Garcia-Recio S, et al.
PLoS ONE, 10(6), e0129661-e0129661 (2015)
Electrochemical and DFT study of the reduction of substituted phenanthrolines
Ferreira H, et al.
Polyhedron, 122, 147-154 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service