Skip to Content
Merck
All Photos(3)

Key Documents

P5708

Sigma-Aldrich

Potassium acetate

BioXtra, ≥99.0%

Synonym(s):

Acetic acid potassium salt, Diuretic salt, KOAc, Potassium ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

pH

7.0-9.0 (25 °C, 98.2 g/L)

solubility

H2O: 5 M, clear, colorless

density

1.57 g/cm3 at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.03%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

agriculture

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium acetate (KAc) is an acetate salt of a weak acid and a strong base. It serves as a buffering agent, maintaining optimal pH levels for enzymatic reactions and protein studies. KAc compatibility with biological systems makes it a preferred choice for the isolation, purification, and analysis of biomolecules. Further, Its ability to stabilize enzymes and maintain optimal conditions in biological assays makes it indispensable in biochemical research. Furthermore, KAc is an excellent choice for chromatography and electrophoresis techniques, facilitating the separation and analysis of biomolecules.

Application


  • Study on the Freeze-Thaw Resistance of Concrete Pavements in Seasonally Frozen Regions.: This study investigates the freeze-thaw resistance of concrete pavements in areas that experience seasonal freezing, emphasizing the role of chemical additives like potassium acetate in enhancing durability (Zhao et al., 2024).

  • Harmonising IV Oxycodone with Paediatric Perioperative Medications: A Compatibility Study Through Y-Type Connectors.: This research explores the compatibility of IV oxycodone with various paediatric perioperative medications, including potassium acetate, to ensure safe administration (Youssef et al., 2024).

  • Gene Expression of Ethanol and Acetate Metabolic Pathways in the Acinetobacter baumannii EmaSR Regulon.: This research examines the metabolic pathways of ethanol and acetate in Acinetobacter baumannii, highlighting the regulatory role of potassium acetate in gene expression (Huang et al., 2024).

Biochem/physiol Actions

Potassium acetate (KAc) buffer aids in the extraction of DNA from various samples including plants and ixodid ticks. It yields a good amount of DNA to test more markers and is highly recommended for use when many markers are to be tested on the same samples. KAc serves as an effective buffer system in chromatography and electrophoresis techniques, for the separation and analysis of biomolecules. In addition, it is also used as an activating agent to prepare activated carbon from waste tea for the adsorption of Acid Blue 25 (AB25) dye.

Features and Benefits

  • The BioXtra product line ensures high-quality products for consistent results.
  • High level of assay ( ≥99.0%), indicating its purity.
  • Powder form for easy handling and use.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Krsek et al.
Journal of microbiological methods, 39(1), 1-16 (1999-12-01)
The efficiency and reproducibility of DNA extraction from soil was tested for variations in lytic and purification treatments and their effect on yield and purity of DNA. The extraction yield was improved by increasing the concentration of EDTA or monovalent
R K Rose et al.
Journal of dental research, 72(1), 78-84 (1993-01-01)
By means of micro-equilibrium dialysis, calcium binding capacities and affinities were measured in three different oral bacteria, and the effects of extracellular polysaccharide, pH, and aggregation were investigated. Binding capacities of 31.0 +/- 2.1 (C. matruchotii), 34.7 +/- 3.7 (S.
Prediction of pH and aw of pork meat by a thermodynamic model: New developments
Ndob A M and Lebert A
Meat Science, 138, 59-67 (2018)
Isolation of oligogalacturonic acids up to DP 20 by preparative high-performance anion-exchange chromatography and pulsed amperometric detection
Hotchkiss Jr A T, et al.
Carbohydrate Research, 334(2), 135- 40 (2001)
An oscillometric detector for capillary electrophoresis
Fracassi da Silva, Jose A and do Lago, Claudimir L
Analytical Chemistry, 70(20) (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service