Skip to Content
Merck
All Photos(1)

Key Documents

P24101

Sigma-Aldrich

Diphenyl ether

ReagentPlus®, 99%

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
Beilstein:
1364620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

>5.86 (25 °C, vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

crystals

autoignition temp.

1144 °F

expl. lim.

1.5 %

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

functional group

phenoxy

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Gene Information

human ... TTR(7276)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diphenyl ether (DPE) is a bridged-phenyl molecule. The conformational analysis of using resonance-enhanced two-photon ionization method DPE has been reported.

Application

Diphenyl ether may be used to synthesize dibenzofuran via intramolecular cyclization in the presence of palladium acetate.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A REMPI investigation of the minimum energy conformations of diphenyl ether.
Paiva ACS, et al.
International Journal of Mass Spectrometry, 221(2), 107-115 (2002)
Palladium-promoted cyclization of diphenyl ether, diphenylamine, and related compounds.
Akermark B, et al.
The Journal of Organic Chemistry, 40(9), 1365-1367 (1975)
Ge-Fei Hao et al.
Journal of computer-aided molecular design, 25(3), 213-222 (2011-01-25)
Protoporphyrinogen oxidase (PPO, EC 1.3.3.4), which has been identified as a significant target for a great family of herbicides with diverse chemical structures, is the last common enzyme responsible for the seventh step in the biosynthetic pathway to heme and
Ariana Beste et al.
The Journal of organic chemistry, 74(7), 2837-2841 (2009-03-06)
Lignin is an abundant natural resource that is a potential source of valuable chemicals. Improved understanding of the pyrolysis of lignin occurs through the study of model compounds for which phenethyl phenyl ether (PhCH(2)CH(2)OPh, PPE) is the simplest example representing
Tony Ly et al.
Analytical chemistry, 80(13), 5059-5064 (2008-05-24)
Selective noncovalent adduct protein probing (SNAPP) mass spectrometry was recently developed to study solution-phase conformations of proteins by exploiting the specific affinity between 18-crown-6 ether (18C6) and lysine side chains. To obtain more detailed information about protein tertiary structure, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service