NIST1974C
Organics in mussel tissue (Mytilus edulis)
NIST® SRM® 1974c
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 jars x 10 g
manufacturer/tradename
NIST®
format
matrix material
General description
Application
- Environmental biomonitoring with mussels: A study assessed the trophic transfer of single-walled carbon nanotubes, using Mytilus edulis as a model organism, to explore the toxicological responses at the base of the food chain, highlighting the role of mussels in environmental biomonitoring (Al-Shaeri et al., 2022).
- Pharmaceutical applications of mussel tissue: Research on the responses of blue mussels to various concentrations of the UV filter ensulizole demonstrated significant findings on the bioactive responses in mussels, useful for understanding bioaccumulation effects relevant to pharmaceutical and environmental monitoring applications (Pham et al., 2022).
- Marine biotechnology research: The investigation into trace elements accumulated in transplanted blue mussels during restoration activities provides insights into environmental and pharmaceutical applications, serving as a model for studying contaminant dynamics and potential health risks in marine biotechnology research (Parolini et al., 2022).
- Biotoxin detection in mussel tissue: A biomarker-based study of the sublethal toxicity of organic UV filters on Mytilus edulis sheds light on the impacts of emerging contaminants, offering crucial data for biotoxin detection and environmental health assessments (Falfushynska et al., 2021).
- Marine biotechnology applications for Mytilus edulis organic extracts: Proteomic analysis of the blue mussel larval shell organic matrix provides a deep understanding of the structural proteins involved, which are critical for developing bioactive materials and applications in marine biotechnology (Carini et al., 2019).
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service