NIST1494
Aliphatic hydrocarbons in 2,2,4-trimethylpentane
NIST® SRM® 1494
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 1.2 mL
manufacturer/tradename
NIST®
format
matrix material
Related Categories
General description
This Standard Reference Material (SRM) is a solution of 20 compounds, including even and odd carbon number aliphatic hydrocarbons from n-decane to n-eicosane, even carbon number aliphatic hydrocarbons from n-eicosane to n-tetratriacontane, and pristine and phytane in 2,2,4-trimethylpentane (iso-octane). This SRM is intended primarily for use in the calibration of chromatographic instrumentation used for the determination of aliphatic hydrocarbons. For more information, please refer to the SDS and COA.
SRM 1494_cert
SRM 1494_SDS
SRM 1494_cert
SRM 1494_SDS
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
n-decane (C10H22), n-undecane (C11H24), n-dodecane (C12H26), n-tridecane (C13H28), n-tetradecane (C14H30), n-pentadecane (C15H32), n-hexadecane (C16H34), n-heptadecane (CH3(CH2)15CH3), pristane (C19H40), n-octadecane (CH3(CH2)16CH3), phytane (C20H42), n-nonadecane (C19H40), n-eicosane (C20H42), n-docosane (C22H46), n-tetracosane (C24H50), n-hexacosane (C26H54), n-octacosane (C28H58), n-triacontane (C30H62), n-dotriacontane (C32H66), n-tetratriacontane (C34H70)
See certificate for values and more details at nist.gov/SRM.
n-decane (C10H22), n-undecane (C11H24), n-dodecane (C12H26), n-tridecane (C13H28), n-tetradecane (C14H30), n-pentadecane (C15H32), n-hexadecane (C16H34), n-heptadecane (CH3(CH2)15CH3), pristane (C19H40), n-octadecane (CH3(CH2)16CH3), phytane (C20H42), n-nonadecane (C19H40), n-eicosane (C20H42), n-docosane (C22H46), n-tetracosane (C24H50), n-hexacosane (C26H54), n-octacosane (C28H58), n-triacontane (C30H62), n-dotriacontane (C32H66), n-tetratriacontane (C34H70)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
10.4 °F - closed cup
Flash Point(C)
-12 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service