Skip to Content
Merck
All Photos(4)

Key Documents

N6522

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

≥98%, BioUltra, from yeast

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
39200202
PubChem Substance ID:
NACRES:
NA.55

biological source

yeast

Quality Level

product line

BioUltra

Assay

≥98%

form

powder

impurities

≤0.1% Insoluble matter

color

white to off-white

solubility

H2O: 0.1 M, clear, colorless to faintly yellow

anion traces

chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • Electrochemical Sensor Development: β-Nicotinamide adenine dinucleotide hydrate is utilized in the development of carbon nanomaterial-based electrochemical sensors and biosensors. These devices are designed for the sensitive detection of pharmaceutical and biological compounds, highlighting its crucial role in enhancing the sensitivity and specificity of biochemical assays (Adhikari et al., 2015).

Biochem/physiol Actions

Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana P Gomes et al.
Cell, 155(7), 1624-1638 (2013-12-24)
Ever since eukaryotes subsumed the bacterial ancestor of mitochondria, the nuclear and mitochondrial genomes have had to closely coordinate their activities, as each encode different subunits of the oxidative phosphorylation (OXPHOS) system. Mitochondrial dysfunction is a hallmark of aging, but
Gaëlle Laurent et al.
Molecular and cellular biology, 33(22), 4552-4561 (2013-09-18)
Sirtuins are a family of protein deacetylases, deacylases, and ADP-ribosyltransferases that regulate life span, control the onset of numerous age-associated diseases, and mediate metabolic homeostasis. We have uncovered a novel role for the mitochondrial sirtuin SIRT4 in the regulation of
Christian Dölle et al.
The FEBS journal, 280(15), 3530-3541 (2013-04-27)
Mitochondrial metabolism is intimately connected to the universal coenzyme NAD. In addition to its role in redox reactions of energy transduction, NAD serves as substrate in regulatory reactions that lead to its degradation. Importantly, all types of the known NAD-consuming
Jeerus Sucharitakul et al.
The Journal of biological chemistry, 288(49), 35210-35221 (2013-10-17)
3-Hydroxybenzoate 6-hydroxylase (3HB6H) from Rhodococcus jostii RHA1 is an NADH-specific flavoprotein monooxygenase that catalyzes the para-hydroxylation of 3-hydroxybenzoate (3HB) to form 2,5-dihydroxybenzoate (2,5-DHB). Based on results from stopped-flow spectrophotometry, the reduced enzyme-3HB complex reacts with oxygen to form a C4a-peroxy
Brian J North et al.
The EMBO journal, 33(13), 1438-1453 (2014-05-16)
Mice overexpressing the mitotic checkpoint kinase gene BubR1 live longer, whereas mice hypomorphic for BubR1 (BubR1(H/H)) live shorter and show signs of accelerated aging. As wild-type mice age, BubR1 levels decline in many tissues, a process that is proposed to

Protocols

To measure alcohol dehydrogenase activity, this assay uses β-nicotinamide adenine dinucleotide phosphate and a continuous spectrophotometric rate determination at 340 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service