Skip to Content
Merck
All Photos(1)

Key Documents

N8410

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide

pkg of 20 mg (per vial)

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2
CAS Number:
Molecular Weight:
663.43
Beilstein:
3584133
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
39200202
PubChem Substance ID:
NACRES:
NA.51

form

powder

Quality Level

packaging

pkg of 20 mg (per vial)

storage temp.

−20°C

SMILES string

NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).
β-Nicotinamide adenine dinucleotide has been used in the master mix for coupled enzyme assay to estimate pectin methylesterase activity.

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide (β-NADH) reduced form is an essential cofactor in all the living organisms. β-NADH functions as a hydride acceptors in biochemical redox reactions. It acts as a cell signaling molecule and is a potent neurotransmitter and neuromodulator in the peripheral nervous system. In brain, it mediates Ca2+ homeostasis, energy metabolism, aging and neuronal death.
Electron acceptor

Other Notes

Packaged based on NAD content as determined by UV-Vis.
This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pectin Methylesterase Activity Assay for Plant Material
Mueller K, et al.
Plant Physiology, 3(17), e894-e894 (2013)
Pectin de-methylesterification and AGP increase promote cell wall remodeling and are required during somatic embryogenesis of Quercus suber
Perez-Perez Y, et al.
Frontiers in Plant Science, 9, e894-e894 (2018)
Irina Lazarova et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 83, 70-78 (2016-10-25)
The aim of this study was to investigate the effect of Asphodeline lutea (L.) Rchb. dry root extract (ALE) administered alone and against carbon tetrachloride (CCl4)-induced liver injury in vitro/in vivo. The dried roots of A. lutea were extracted with
Release, neuronal effects and removal of extracellular beta-nicotinamide adenine dinucleotide beta-NAD+ in the rat brain
Durnin L, et al.
The European Journal of Neuroscience, 35(3), 423-435 (2012)
The biosynthesis of nicotinamide adenine dinucleotides in bacteria
Begley TP, et al.
Vitamins and Hormones, 61, 103-119 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service