M8911
Methyl 4-hydroxybenzoate
analytical standard
Synonym(s):
p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
509801
EC Number:
MDL number:
UNSPSC Code:
51452201
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
grade
analytical standard
Quality Level
Assay
98.0-102.0%
form
solid
packaging
vial of 200 mg
mp
125-128 °C (lit.)
SMILES string
COC(=O)c1ccc(O)cc1
InChI
1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
InChI key
LXCFILQKKLGQFO-UHFFFAOYSA-N
Gene Information
rat ... Ar(24208)
Looking for similar products? Visit Product Comparison Guide
Application
Methyl 4-hydroxybenzoate has been used as an internal standard for the determination of trans-10-hydroxy-2-decenoic acid (10-HDA).
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Supplied in amber screw-cap vials
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
334.4 °F
Flash Point(C)
168 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Optimized determination method for trans-10-hydroxy-2-decenoic acid content in royal jelly by high-performance liquid chromatography with an internal standard
Zhou J, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 90(1), 244-249 (2007)
Comparison of UPLC and HPLC for determination of trans-10-hydroxy-2-decenoic acid content in royal jelly by ultrasound-assisted extraction with internal standard
Zhou J, et al.
Chromatographia, 66(3-4), 185-190 (2007)
Kyung-Jin Min et al.
Aging cell, 7(2), 199-206 (2008-01-29)
Reduced insulin/insulin-like growth factor (IGF) signaling may be a natural way for the reduction of dietary nutrients to extend lifespan. While evidence challenging this hypothesis is accumulating with Caenorhabditis elegans, for Drosophila melanogaster it is still thought that insulin/IGF and
Ismail I Hewala et al.
Talanta, 88, 623-630 (2012-01-24)
Simple methods for HPTLC peak purity assessment and identification of the HPTLC peaks were presented. The spectrodensitograms - selected at different time intervals across the elution time of the HPTLC peak - were extracted and digital algorithms for manipulating the
Gabriela Oliveira et al.
International journal of pharmaceutics, 435(1), 38-49 (2012-05-29)
Simple topical formulations which include volatile components, such as gels or sprays, are appealing from a cosmetic perspective. However, complex formulation effects may result from the use of volatile excipients in topical formulations, particularly when applied at clinically relevant doses
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service