Skip to Content
Merck
All Photos(1)

Key Documents

M3516

Sigma-Aldrich

Mineral oil

suitable for preparation of Nujol mulls for infrared spectroscopy, light oil

Synonym(s):

White mineral oil (petroleum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

form

light oil

Quality Level

technique(s)

PCR: suitable

refractive index

n20/D 1.467 (lit.)

density

0.84 g/mL at 25 °C (lit.)

suitability

suitable for preparation of Nujol mulls for infrared spectroscopy

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mineral oils are complex mixtures of straight and branched chain paraffinic, naphthenic and aromatic hydrocarbons refined from petroleum crude oils. Mineral oil with a carbon number of 15 to 25 is categorized as light oil and 25 to 50 as heavy oil. Mineral oil is commonly used in food, cosmetics and pharmaceutical industries especially in skin care products. Mineral oil is used as overlaying agent in polymerase chain reaction (PCR) and electrophoretic techniques to avoid evaporation.

Application

Mineral oil has been used:
  • to overlay the samples to avoid evaporation in denaturing gel PCR reaction mix
  • in emulsion preparation for emulsion PCR

Packaging

6 mL size is supplied in plastic dropper bottle.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroserpin Portland (Ser52Arg) is trapped as an inactive intermediate that rapidly forms polymers: implications for the epilepsy seen in the dementia FENIB
Belorgey D, et al.
European Journal of Biochemistry, 271(16), 3360-3367 (2004)
Intraspecific variation of Rhizoctonia solani AG 3 isolates recovered from potato fields in Central Iran and South Australia
Balali GR, et al.
Mycopathologia, 163(2), 105-115 (2007)
Bisulfite genomic sequencing: systematic investigation of critical experimental parameters
Grunau C, et al.
Nucleic Acids Research, 29(13), e65-e65 (2001)
Hideyuki Watanabe et al.
Biomedical optics express, 2(8), 2279-2287 (2011-08-13)
Here, we report in vivo 3-D visualization of the layered organization of a rat olfactory bulb (OB) by a swept source optical coherence tomography (SS-OCT). The SS-OCT operates at a wavelength of 1334 nm with respective theoretical depth and lateral
A review on the extensive skin benefits of mineral oil
Rawlings AV and Lombard KJ
International Journal of Cosmetic Science, 34(6), 511-518 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service