M0678
Middlebrook OADC Growth Supplement
Enrichment supplement recommended for isolation and cultivation of Mycobacteria, suitable for microbiology
Synonym(s):
Middlebrook Oleic Albumin Dextrose Catalase Growth Supplement, Oleic Albumin Dextrose Catalase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171614
NACRES:
NA.85
Recommended Products
Quality Level
sterility
sterile (Filtered and Aseptic Handled)
form
liquid
shelf life
limited shelf life, expiry date on the label
packaging
vial of 50 mL
application(s)
clinical testing
environmental
food and beverages
life science and biopharma
veterinary
microbiology
storage temp.
2-8°C
suitability
mycobacteria
General description
Middlebrook OADC Growth Supplement is used with the Middlebrook Agar, that allows the isolation and cultivation of a wide variety of Mycobacteria. Middlebrook OADC Growth Supplement contains oleic acid, bovine albumin, sodium chloride, dextrose, and catalase. Oleic acid and other long chain fatty acids are essential for the metabolism of Mycobacteria. Some free fatty acids are toxic to Mycobacteria, but albumin binds to those fatty acids and prevents toxic action on Mycobacteria. Dextrose is the fermentable carbohydrate and an energy source whereas catalase destroys toxic peroxides that may be present in the medium.
Application
Middlebrook OADC growth supplement has been used as a supplement in 7H9 liquid medium to culture mycobacteria from clinical samples, environmental, food and beverages.
Components
Ingredients: (per vial)
Bovine Albumin Fraction V 2.50 g
Dextrose 1.00 g
Catalase 0.002 g
Oleic acid 0.025 g
Sodium chloride 0.425 g
Distilled water 50 mL
Bovine Albumin Fraction V 2.50 g
Dextrose 1.00 g
Catalase 0.002 g
Oleic acid 0.025 g
Sodium chloride 0.425 g
Distilled water 50 mL
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Detecting phenotypically resistant Mycobacterium tuberculosis using wavelength modulated Raman spectroscopy
Antibiotic resistance protocols, 41-50 (2018)
Recombinant BCG Vaccines Reduce Pneumovirus-Caused Airway Pathology by Inducing Protective Humoral Immunity
Soto JA, et al.
Frontiers in immunology, 9 (2018)
Recombinant BCG Vaccines Reduce Pneumovirus-Caused Airway Pathology by Inducing Protective Humoral Immunity
Soto JA, et al.
Frontiers in Immunology, 9 (2018)
Shuxin Liang et al.
Autophagy, 16(8), 1453-1467 (2019-11-13)
BAG2 (BCL2 associated athanogene 2) is associated with cell fate determination in response to various pathological conditions. However, the effects of BAG2 on M. tuberculosis-induced endoplasmic reticulum (ER) stress remain elusive. Herein, we report that M. tuberculosis infection of macrophages
Susanta Pahari et al.
Autophagy, 16(6), 1021-1043 (2019-08-30)
Host-directed therapies are gaining considerable impetus because of the emergence of drug-resistant strains of pathogens due to antibiotic therapy. Therefore, there is an urgent need to exploit alternative and novel strategies directed at host molecules to successfully restrict infections. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service