Skip to Content
Merck
All Photos(1)

Key Documents

L0380000

Levamisole hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
Beilstein:
4358988
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levamisole

manufacturer/tradename

EDQM

mp

228-230 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.C1CN2C[C@@H](N=C2S1)c3ccccc3

InChI

1S/C11H12N2S.ClH/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10;/h1-5,10H,6-8H2;1H/t10-;/m1./s1

InChI key

LAZPBGZRMVRFKY-HNCPQSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levamisole hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingxin Li et al.
Annals of hematology, 92(9), 1239-1247 (2013-04-27)
The appropriate management of patients with moderate aplastic anemia (mAA) remains to be unclear and controversial. A cohort of 118 patients with mAA received a novel immunosuppressive strategy of cyclosporine alternately combined with levamisole (CSA and LMS regimen), which included
M P Myklebust et al.
British journal of cancer, 107(10), 1684-1691 (2012-10-27)
The aim of this study was to investigate the value of the cyclin D1 isoforms D1a and D1b as prognostic factors and their relevance as predictors of response to adjuvant chemotherapy with 5-fluorouracil and levamisole (5-FU/LEV) in colorectal cancer (CRC).
Ana Mbokeleng Tsotetsi et al.
Tropical animal health and production, 45(3), 751-761 (2012-10-16)
The present study was conducted to determine the prevalence and distribution of gastrointestinal helminths, to detect the presence of anthelmintic resistance in livestock from small-scale farms and to determine the level of helminthosis awareness among small-scale farmers in Gauteng Province
Kelly R Magliocca et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 71(3), 487-492 (2013-01-10)
Systemic complications of levamisole-adulterated cocaine (LAC) use have recently been described. The objective of this review is to increase awareness of these manifestations among oral and maxillofacial surgeons. LAC exposure through inhalation, nasal insufflation, or injection can induce cutaneous vasculopathy
David S Lindsay et al.
Veterinary parasitology, 196(1-2), 21-23 (2013-03-26)
Decoquinate is a quinolone anticoccidial approved for use in the prevention of intestinal coccidiosis in farm animals. This compound has good activity against Toxoplasma gondii and Neospora caninum in cell cultures. The drug acts on the parasites' mitochondria. The activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service