Skip to Content
Merck
All Photos(1)

Key Documents

H4777

Sigma-Aldrich

Heparan sulfate proteoglycan

≥400 μg/mL glycosaminoglycan

Synonym(s):

HSPG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

mouse (Engelbreth-Holm-Swarm mouse sarcoma basement membrane)

Quality Level

description

For cell culture use.
Isolated from basement membrane of Engelbreth-Holm-Swarm mouse sarcoma.
Solution in 50 mM Tris HCl, 150 mM NaCl, 1 mM EDTA, 0.1 mM PMSF, pH 7.4, containing ≥400 μg protein per ml.

form

solution

mol wt

≥400 kDa

packaging

pkg of 0.1 mg

concentration

≥400 μg/mL glycosaminoglycan

technique(s)

cell culture | mammalian: suitable

impurities

≥100 μg/mL Uronic acid

NCBI accession no.

UniProt accession no.

Binding Specificity

Peptide Source: Collagen Type IV

Peptide Source: Fibronectin

Peptide Source: Laminin

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Hspg2(15530)

General description

Heparan sulfate proteoglycan (HSPG), a glycoprotein, is an integral part of the basement membrane. It is composed of a core protein covalently bound to heparan sulfate chains. There are three groups of HSPGs: membrane HSPGs, secreted extracellular matrix HSPGs, and the secretory vesicle proteoglycan.

Application

Heparan sulfate proteoglycan has been used:

  • in phosphate buffered saline (PBS) and added to fresh citrated blood samples to achieve trauma-equivalent concentrations in a research study
  • solid phase binding assay
  • for fabrication of extracellular matrix (ECM) microarray slides

Biochem/physiol Actions

Heparan sulfate proteoglycans (HSPGs) have multiple functions in various biological processes. They contribute to basement membrane structure and provide a matrix for cell migration. HSPGs in secretory vesicles, such as serglycin, play a role in packaging granular contents, maintaining proteases activity, and regulating biological activities after secretion. HSPGs can bind and protect cytokines, growth factors, and chemokines, facilitating the formation of gradients essential for cell specification and leukocyte recruitment. They act as receptors for proteases and protease inhibitors, regulate cell-ECM attachment, cell-cell interactions, and cell motility, and can cooperate with growth factor receptors. HSPGs also function as endocytic receptors for ligand clearance and are involved in lipoprotein metabolism and morphogen gradient formation. It also acts as a docking molecule for matrilysin and other matrix metalloproteinases. Heparan Sulfate promotes the attachment of cells in culture and induces high-affinity binding of FGF-basic to cells deficient in heparin sulfate and soluble FGF receptors at a concentration of 10-100 ng/mL.

Components

Heparan sulfate proteoglycan is a large biomolecule composed of a core protein covalently bound to heparin sulfate chains that is an integral part of the basement membrane. The number of polysaccharide chains and the size of the core protein are variable according to the source.

Caution

It is recommended to store this product at 20°C, where it will retain its function for 3 years

Preparation Note

This product is isolated from the basement membrane of Engelbreth-Holm-Swarm mouse sarcoma in a solution containing 50 mM Tris HCl at pH 7.4, 150 mM NaCl, 1 mM EDTA, and 0.1 mM PMSF.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heparan sulfate proteoglycans
Stephane Sarrazin , William C Lamanna, Jeffrey D Esko
Cold Spring Harbor Perspectives in Biology, 3(7) (2011)
C Battaglia et al.
European journal of biochemistry, 208(2), 359-366 (1992-09-01)
A large, low-density form of heparan sulfate proteoglycan was isolated from the Engelbreth-Holm-Swarm (EHS) tumor and demonstrated to bind in immobilized-ligand assays to laminin fragment E3, collagen type IV, fibronectin and nidogen. The first three ligands mainly recognize the heparan
Glycocalyx components affect platelet function, whole blood coagulation, and fibrinolysis: an in vitro study suggesting a link to trauma-induced coagulopathy
Martin W Britten, et al.
BMC anesthesiology, 21(1), 83-83 (2021)
Pongsak Khunrae et al.
Journal of molecular biology, 397(3), 826-834 (2010-01-30)
Plasmodium falciparum malaria remains one of the world's leading causes of human suffering and poverty. Each year, the disease takes 1-3 million lives, mainly in sub-Saharan Africa. The adhesion of infected erythrocytes (IEs) to vascular endothelium or placenta is the
Feng Long et al.
Bone, 142, 115686-115686 (2020-10-16)
Endochondral ossification is the major process of long bone formation, and chondrogenesis is the final step of this process. Several studies have indicated that bone morphogenetic proteins (BMPs) are required for chondrogenesis and regulate multiple growth plate features. Abnormal BMP

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service