Skip to Content
Merck
All Photos(2)

Key Documents

F8880

Supelco

Fluocinolone acetonide

analytical standard

Synonym(s):

6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Sinalar, Synandone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30F2O6
CAS Number:
Molecular Weight:
452.49
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

267-269 °C (lit.)

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

InChI

1S/C24H30F2O6/c1-20(2)31-19-9-13-14-8-16(25)15-7-12(28)5-6-21(15,3)23(14,26)17(29)10-22(13,4)24(19,32-20)18(30)11-27/h5-7,13-14,16-17,19,27,29H,8-11H2,1-4H3/t13-,14-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

FEBLZLNTKCEFIT-VSXGLTOVSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Fluocinolone acetonide may be used as a reference standard in the analysis of fluocinolone acetonide in samples of soil via pressurized liquid extraction (PLE) and solid-phase extraction (SPE) followed by liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical method for the determination of trace levels of steroid hormones and corticosteroids in soil, based on PLE/SPE/LC-MS/MS.
Gineys N, et al.
Analytical and Bioanalytical Chemistry, 397(6), 2295-2302 (2010)
John H Kempen et al.
Ophthalmology, 120(9), 1852-1859 (2013-05-28)
To evaluate agreement between fluorescein angiography (FA) and optical coherence tomography (OCT) results for diagnosis of macular edema in patients with uveitis. Multicenter cross-sectional study. Four hundred seventy-nine eyes with uveitis from 255 patients. The macular status of dilated eyes
Soraya Rofagha et al.
Ocular immunology and inflammation, 21(1), 77-78 (2013-01-18)
To describe a case of spontaneous dissociation of a fluocinolone acetonide implant (Retisert) 5 years following implantation. Case report. A patient with chronic uveitis underwent placement of a fluocinolone acetonide implant in the right eye in 2006. The patient's inflammation
Bryn M Burkholder et al.
Retina (Philadelphia, Pa.), 33(8), 1684-1693 (2013-04-04)
To evaluate outcomes after placement of fluocinolone acetonide (FA) implants in eyes with birdshot chorioretinitis and to compare these outcomes with eyes with posterior and panuveitis. This is a retrospective cohort study of 48 eyes from patients with posterior and
Oliver Comyn et al.
Current opinion in ophthalmology, 24(3), 248-254 (2013-03-23)
Three long-acting corticosteroid implants are now available for the treatment of retinal disease, offering control of macular edema and inflammation for between 6 months and up to 3 years. This review evaluates their efficacy and side-effect profile in comparison with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service