Skip to Content
Merck
All Photos(3)

Key Documents

D9684

Sigma-Aldrich

Dextromethorphan hydrobromide

meets USP testing specifications

Synonym(s):

Dextromethorphan hydrobromide monohydrate, (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32
Beilstein:
6453793
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

98.0-102.0% anhydrous basis

form

solid

SMILES string

Br[H].[H]O[H].[H][C@]12CCCC[C@]13CCN(C)[C@H]2Cc4ccc(OC)cc34

InChI

1S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H;1H2/t15-,17+,18+;;/m1../s1

InChI key

STTADZBLEUMJRG-IKNOHUQMSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinn-Jong Jiang et al.
Microcirculation (New York, N.Y. : 1994), 20(2), 190-201 (2012-11-13)
This study examines the effect of Dextromethorphan (d-3-methoxy-17-methylmorphinan; DXM), a commonly used cough-suppressing drug, on the expression of VCAM-1 and ICAM-1 in human umbilical vein endothelial cells (HUVECs) stimulated with lipopolysaccharide (LPS). The effect of DXM on expression of cell
Ewa Tomczak et al.
Archiwum medycyny sadowej i kryminologii, 62(3), 197-202 (2013-05-09)
The study highlighted the problem of intoxication using substances and/or preparations, to which nowadays young people have unrestricted access. Based on the case developed in the Department of Forensic Medicine of the Medical University of Gdansk, our team members were
Fixed drug eruption due to dextromethorphan with tolerance to other opioids.
G Davila-Fernández et al.
Journal of investigational allergology & clinical immunology, 23(4), 281-282 (2013-08-24)
K N Chi et al.
Cancer chemotherapy and pharmacology, 71(1), 237-244 (2012-10-16)
To assess the effect of abiraterone acetate plus prednisone on the pharmacokinetics of dextromethorphan HBr (CYP2D6 substrate) and theophylline (CYP1A2 substrate) in patients with metastatic castration-resistant prostate cancer (mCRPC). Men with progressive metastatic mCRPC who failed gonadotropin-releasing hormone therapy and
F L Opdam et al.
Cancer chemotherapy and pharmacology, 71(3), 593-601 (2012-12-12)
Adjuvant therapy with tamoxifen significantly reduces breast cancer recurrence and mortality in estrogen receptor positive disease. CYP2D6 is the main enzyme involved in the activation of the prodrug tamoxifen into the anti-estrogen endoxifen. Endoxifen is thought to be a main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service