Skip to Content
Merck
All Photos(5)

Key Documents

D8255

Sigma-Aldrich

1,4-Dithioerythritol

≥99.0%

Synonym(s):

erythro-1,4-Dimercapto-2,3-butanediol, erythro-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719756
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0%

form

powder

reaction suitability

reagent type: reductant

pH

4.5-6.5 (25 °C, 15.4 g/L)

mp

82-84 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for maintaining −SH groups in the reduced state; quantitatively reduces disulfides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin M Cleary et al.
eLife, 9 (2020-09-15)
Respiratory chemoreceptors regulate breathing in response to changes in tissue CO2/H+. Blood flow is a fundamental determinant of tissue CO2/H+, yet little is known regarding how regulation of vascular tone in chemoreceptor regions contributes to respiratory behavior. Previously, we showed
Christopher J Garland et al.
Science signaling, 10(486) (2017-07-06)
Vascular smooth muscle contraction is suppressed by feedback dilation mediated by the endothelium. In skeletal muscle arterioles, this feedback can be activated by Ca2+ signals passing from smooth muscle through gap junctions to endothelial cells, which protrude through holes in
Muzamil M Z Noorani et al.
Frontiers in physiology, 2, 42-42 (2011-08-04)
Transient receptor potential (TRP) C1 and C3 (TRPC1 and TRPC3) are expressed in vascular smooth muscle cells and are thought to be involved in vascular contractility. In the present study, we determined the effect of systemic hypertension on TRPC1/TRPC3 channel
Vishal R Yadav et al.
American journal of physiology. Lung cellular and molecular physiology, 314(5), L724-L735 (2018-02-02)
Hypoxia-induced pulmonary vasoconstriction (HPV) is attributed to an increase in intracellular Ca2+ concentration ([Ca2+]i) in pulmonary artery smooth muscle cells (PASMCs). We have reported that phospholipase C-γ1 (PLCγ1) plays a significant role in the hypoxia-induced increase in [Ca2+]i in PASMCs
Stephen Shrum et al.
Biomolecules, 9(12) (2019-12-11)
Kidneys from deceased donors used for transplantation are placed in cold storage (CS) solution during the search for a matched recipient. However, CS causes mitochondrial injury, which may exacerbate renal graft dysfunction. Here, we explored whether adding NS11021, an activator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service