Skip to Content
Merck
All Photos(1)

Key Documents

D0600000

Deslanoside

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Deacetyllanatoside C, Desacetyllanatoside C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H74O19
CAS Number:
Molecular Weight:
943.08
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

deslanoside

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C47H74O19/c1-20-41(64-36-16-30(50)42(21(2)60-36)65-37-17-31(51)43(22(3)61-37)66-44-40(56)39(55)38(54)32(18-48)63-44)29(49)15-35(59-20)62-25-8-10-45(4)24(13-25)6-7-27-28(45)14-33(52)46(5)26(9-11-47(27,46)57)23-12-34(53)58-19-23/h12,20-22,24-33,35-44,48-52,54-57H,6-11,13-19H2,1-5H3/t20-,21-,22-,24-,25+,26-,27-,28+,29+,30+,31+,32-,33-,35+,36+,37+,38-,39+,40-,41-,42-,43-,44+,45+,46+,47+/m1/s1

InChI key

OBATZBGFDSVCJD-LALPQLPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Deslanoside EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Histoenzymatic studies of the kidneys of albino rats after experimental administration of serpasil and deslanoside].
A Zarebska
Patologia polska, 36(2), 200-207 (1985-04-01)
[Positive inotropic effect of digitalis on the left ventricular systolic time interval in patients with primary dilated cardiomyopathy observed during increased heart ventricle load].
J P Dubiel et al.
Przeglad lekarski, 43(6), 433-438 (1986-01-01)
Topochemical examinations of the lungs of the rat after combined administration of serpasil and deslanatoside. I. Histochemical examinations.
E A Jedrzejewska
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 42, 133-138 (1987-01-01)
A Fronczak et al.
Polish journal of pharmacology and pharmacy, 41(6), 597-609 (1989-11-01)
4'-epidoxorubicin (4'-epiDXR) was compared with doxorubicin (DXR) for myocardial effects in rabbits. Histologically, both drugs when given for six weeks induced a high incidence of diffused myocardial damage. The myocardial damage was less evident in rabbits treated with 4'-epiDXR than
Y Fujii et al.
Journal of chromatographic science, 28(6), 288-291 (1990-06-01)
A micro high-performance liquid chromatographic (micro-HPLC) procedure for the assay of digitoxin tablets and deslanoside injections has been developed. Micro-HPLC is performed on an ODS micro column, with acetonitrile-methanol-water (10:20:17) for digitoxin tablets and acetonitrile-water (21:70) for deslanoside injections. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service