
Deslanoside

Synonym(s):
Deacetyllanatoside C, Desacetyllanatoside C
Empirical Formula (Hill Notation):
C47H74O19
CAS Number:
Molecular Weight:
943.08
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing