Skip to Content
Merck
All Photos(1)

Key Documents

D0400000

Desoxycortone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Deoxycorticosterone acetate, 11-Deoxycorticosterone acetate, 21-Acetoxy-4-pregnene-3,20-dione, 21-Acetoxyprogesterone, 21-Hydroxy-4-pregnene-3,20-dione 21-acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnen-21-ol-3,20-dione 21-acetate, Cortexone acetate, DOC acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
Beilstein:
2570798
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

desoxycortone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@]([H])(CC[C@@]24[H])C(=O)COC(C)=O

InChI

1S/C23H32O4/c1-14(24)27-13-21(26)20-7-6-18-17-5-4-15-12-16(25)8-10-22(15,2)19(17)9-11-23(18,20)3/h12,17-20H,4-11,13H2,1-3H3/t17-,18-,19-,20+,22-,23-/m0/s1

InChI key

VPGRYOFKCNULNK-ACXQXYJUSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Desoxycortone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Vandorpe et al.
Canadian journal of physiology and pharmacology, 69(11), 1784-1788 (1991-11-01)
Rebound metabolic alkalosis is a transient alkalemia that is seen during recovery from NH4Cl-induced metabolic acidosis. The persistent elevation of plasma bicarbonate concentration is the result of continuing excretion of net acid by the kidney. Bicarbonate transport by inner medullary
Jiang Xu et al.
American journal of physiology. Heart and circulatory physiology, 299(5), H1328-H1338 (2010-09-14)
Angiotensin II (ANG II) contributes to hypertension, cardiac hypertrophy, fibrosis, and dysfunction; however, it is difficult to separate the cardiac effect of ANG II from its hemodynamic action in vivo. To overcome the limitations, we used transgenic mice with cardiac-specific
Carrie A Northcott et al.
Experimental biology and medicine (Maywood, N.J.), 237(1), 99-110 (2012-01-10)
One challenge in understanding the polygenic disease of hypertension is elucidating the genes involved and defining responses to environmental factors. Many studies focus on animal models of hypertension; however, this does not necessarily extrapolate to humans. Current technology and cost
Saad Lahlou et al.
Fundamental & clinical pharmacology, 17(3), 323-330 (2003-06-14)
The present study investigated the hypotensive responses to intravenous (i.v.) treatment with the essential oil of Alpinia zerumbet (EOAZ) and its main constituent, terpinen-4-ol (Trp-4-ol), in the experimental model of deoxycorticosterone-acetate (DOCA)-salt hypertensive rat. In both DOCA-salt hypertensive and uninephrectomized
Riccardo Candido et al.
American journal of hypertension, 18(6), 844-850 (2005-06-01)
The aim of the present study was to compare glomerular permeability alterations associated with experimental hypertension models known to have different effects on the circulating renin-angiotensin system (RAS). Five groups, 10 animals each, were studied. One group served as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service