Skip to Content
Merck
All Photos(1)

Key Documents

PHR1026

Supelco

Ranitidine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N, N Dimethyl-5-[2-(1-methylamine-2-nitrovinyl)-ethylthiomethyl]furfurylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N4O3S · HCl
CAS Number:
Molecular Weight:
350.86
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 471
traceable to Ph. Eur. R0150000
traceable to USP 1598405

API family

ranitidine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].CN\C(NCCSCc1ccc(CN(C)C)o1)=C\[N+]([O-])=O

InChI

1S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15H,6-8,10H2,1-3H3;1H/b13-9-;

InChI key

GGWBHVILAJZWKJ-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ranitidine hydrochloride is a H2-receptor antagonist, widely used for the management of hypersecretory conditions and for the treatment of duodenal ulcer.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ranitidine hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using titrimetry and visible spectrophotometry techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

H2 histamine receptor antagonist; anti-ulcer agent.
H2 histamine receptor antagonist; anti-ulcer agent.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5630 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of ranitidine hydrochloride based on the reaction with p-dimethylaminobenzaldehyde
Narayana B, et al.
Eurasian Journal of Analytical Chemistry, 5(1), 63-72 (2010)
Peter J Kahrilas et al.
Chest, 143(3), 605-612 (2012-11-03)
Epidemiologic and physiologic studies suggest an association between gastroesophageal reflux disease (GERD) and chronic cough. However, the benefit of antireflux therapy for chronic cough remains unclear, with most relevant trials reporting negative findings. This systematic review aimed to reevaluate the
M Boyce et al.
Alimentary pharmacology & therapeutics, 36(2), 181-189 (2012-05-23)
Nonclinical studies have shown netazepide (YF476) to be a potent, selective, competitive and orally active gastrin receptor antagonist. To administer to humans for the first time single oral doses of netazepide, to assess their tolerability, safety, pharmacokinetics and effect on
Elvira Escribano et al.
International journal of pharmaceutics, 436(1-2), 472-477 (2012-07-21)
The aim of the present work was to study the intestinal permeabilities (P(eff)) of five model drugs: furosemide, piroxicam, naproxen, ranitidine and amoxicillin in the in situ intestinal perfusion technique in mice and compare them with corresponding rat and human
Michael J Dolton et al.
Antimicrobial agents and chemotherapy, 56(11), 5503-5510 (2012-08-15)
Posaconazole has an important role in the prophylaxis and salvage treatment of invasive fungal infections (IFIs), although poor and variable bioavailability remains an important clinical concern. Therapeutic drug monitoring of posaconazole concentrations has remained contentious, with the use of relatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service