Skip to Content
Merck
All Photos(1)

Key Documents

C0470000

Carbocisteine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

S-Carboxymethyl-L-cysteine, 3-carboxymethylthio-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4S
CAS Number:
Molecular Weight:
179.19
Beilstein:
1725012
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carbocisteine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N[C@@H](CSCC(O)=O)C(O)=O

InChI

1S/C5H9NO4S/c6-3(5(9)10)1-11-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

GBFLZEXEOZUWRN-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbocisteine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boontarika Boonyapiwat et al.
The Journal of pharmacy and pharmacology, 63(4), 558-564 (2011-03-16)
The aim of this investigation was to provide in-vitro enzyme kinetic data to support the hypothesis that the in-vivo heterozygous dominant phenotype for phenylalanine monooxygenase (hPAH) was responsible for the S-oxidation polymorphism in the metabolism of S-carboxymethyl-l-cysteine reported in humans.
Masayuki Hanaoka et al.
Chest, 139(5), 1101-1108 (2010-09-18)
Chronic inflammation, imbalance of proteolytic and antiproteolytic activities, oxidative stress, and apoptosis of lung structural cells contribute to the pathogenesis of COPD. There is increasing evidence that carboxymethylcysteine (also known as carbocisteine) (CMC), which is commonly used for its mucoactive
[Notes on syrups and dry syrups of ambroxol hydrochloride and L-carbocysteine].
Naoto Watanabe et al.
Arerugi = [Allergy], 61(5), 659-661 (2012-08-29)
Matthew Thomas Hardison et al.
Frontiers in bioscience (Elite edition), 4, 2402-2409 (2012-06-02)
Several chronic lung diseases have been linked to cigarette smoking (Chronic Obstructive Pulmonary Disease (COPD), and cancer are associated with increased tobacco use). We recently described a collagen fragment, proline-glycine-proline (PGP), chemotactic for neutrophils, that appears to play a role
Yuji Ishibashi et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 132(6), 699-704 (2012-06-13)
Human bronchial mucins, such as MUC5AC, have traditionally been defined as a family of high-molecular weight glycoproteins. Changes in the contents of sugar chains on MUC5AC are among the fundamental features in inflammatory respiratory disease. The changes have been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service