Skip to Content
Merck
All Photos(6)

Key Documents

A26357

Sigma-Aldrich

Adipic acid

99%

Synonym(s):

ADI-PURE® LGA adipic acid, ADI-PURE® high purity adipic acid, Hexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1209788
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021711
PubChem Substance ID:
NACRES:
NA.21

vapor density

5 (vs air)

Quality Level

vapor pressure

1 mmHg ( 159.5 °C)

Assay

99%

form

solid

autoignition temp.

788 °F

bp

265 °C/100 mmHg (lit.)

mp

151-154 °C (lit.)

solubility

H2O: soluble 23 g/L at 25 °C

SMILES string

OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adipic acid is a straight-chain aliphatic dicarboxylic acid, commonly used in the manufacturing of nylon-6,6 and plasticizers. Conventionally it was manufactured from petrochemicals but in recent days it can be synthesized from renewable substrates by means of biological methods.

Application

Adipic acid can be used as a starting material in the preparation of:
  • Aliphatic polyesters by reacting with ethyleneglycol/1,3 propyleneglycol/1,4-butanediol using inorganic acid as a catalyst.
  • Cyclopentanone using a weak base such as Na2CO3.
  • Linear polybutylene adipate (PBA) having carboxylic acids at the terminals by reacting with 1,4-butanediol.

Legal Information

ADI-PURE is a registered trademark of Invista North America S.a.r.l.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of aliphatic polyesters by polycondensation using inorganic acid as catalyst
Sokolsky-Papkov M, et al.
Polymers For Advanced Technologies, 22(5), 502-511 (2011)
Biological production of adipic acid from renewable substrates: Current and future methods
Deng Y, et al.
Biochemical Engineering Journal, 105, 16-26 (2016)
Ketonic decarboxylation catalysed by weak bases and its application to an optically pure substrate
Renz M and Corma A
European Journal of Organic Chemistry, 2004(9), 2036-2039 (2004)
Synthesis and characterization of high molecular weight branched PBA
Han Y, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 39(13), 2143-2150 (2001)
Migration resistant polymeric plasticizer for poly (vinyl chloride)
Lindstrom A and Hakkarainen M
Journal of Applied Polymer Science, 104(4), 2458-2467 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service