Skip to Content
Merck
All Photos(1)

Key Documents

BCR270

Triphenylene

BCR®, certified reference material

Synonym(s):

9,10-Benzophenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1342908
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

438 °C (lit.)

mp

195-198 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)c3ccccc3c4ccccc24

InChI

1S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12H

InChI key

SLGBZMMZGDRARJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR270

Legal Information

BCR is a registered trademark of European Commission

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ozcan Kocyigit et al.
Journal of hazardous materials, 183(1-3), 334-340 (2010-08-10)
We presented the synthesis of a novel Schiff base and its complexation properties with some transition metal ions in this work. For this, the 2,3,4,6,7,10,11 hexahydroxytriphenylene (HHTP) as starting material was synthesized according to known procedure. Moreover, the 2-((2-chloroethylimino)methyl) phenol
Lucas A Haverkate et al.
The journal of physical chemistry. B, 116(43), 13098-13105 (2012-10-05)
Discotic liquid crystalline (DLC) charge transfer (CT) complexes, which combine visible light absorption with rapid charge transfer characteristics within the CT complex, can have a great potential for photovoltaic applications when they can be made to self-assemble in a bulk
Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 41(37), 11413-11418 (2012-08-15)
The binding behavior of triphenylene based copper ensemble prepared in situ has been investigated toward various anions (F(-), Cl(-), Br(-), I(-), CH(3)COO(-), H(2)PO(4)(-), NO(3)(-), OH(-), ClO(4)(-), CN(-), CO(3)(-) and SO(4)(-)) by UV-vis and fluorescence spectroscopy. Among various anions tested, 1-Cu(2+)
Fabián Vaca Chávez et al.
The journal of physical chemistry. B, 116(8), 2339-2346 (2012-02-23)
The Larmor frequency and temperature dependence of the proton nuclear magnetic resonance (NMR) spin-lattice relaxation time was measured in the isotropic and columnar phases of both chain-end fluorinated triphenylene disk-like and fully hydrogenated molecules. In the columnar phase, the results
Mahuya Bagui et al.
The journal of physical chemistry. A, 115(9), 1579-1592 (2011-02-12)
A donor-acceptor charge transfer system based on two discotic mesogens has been synthesized. The donor is either a triphenylene (POG0) or a triphenylene-based conjugated dendron (POG1), while the acceptor is a perylene diimide (PDI) core. The donors are covalently linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service