Skip to Content
Merck
All Photos(1)

Key Documents

B1116000

Biotin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

biotin

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

231-233 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Biotin is a water-soluble vitamin B, which is involved in the synthesis of fatty acids and in gluconeogenesis. It is found in plants and animals. Biotin deficiency causes retardation in growth, alopecia and neurological impairment in humans and animals.

Application

Biotin may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biotin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 16(8), 5631-5632 (2018)
Agar plate method using Lactobacillus plantarum for biotin determination in serum and urine
FUKUI T, et al.
J. Nutr. Sci. Vitaminol., 40(5), 491-498 (1994)
Factitious Graves disease due to biotin immunoassay interference-a case and review of the literature
Elston MS, et al.
The Journal of clinical endocrinology and metabolism, 101(9), 3251-3255 (2016)
Magnetic resonance imaging assays for dimethyl sulfoxide effect on cancer vasculature
Cyran CC, et al.
Investigative Radiology, 43(5), 298-298 (2008)
Liquid chromatographic determination of biotin in multivitamin-multimineral tablets.
Ekpe AE and Hazen C
Journal of Pharmaceutical and Biomedical Analysis, 16(8), 1311-1315 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service