Skip to Content
Merck
All Photos(1)

Key Documents

B1115000

Bezafibrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClNO4
CAS Number:
Molecular Weight:
361.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bezafibrate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1)C(O)=O

InChI

1S/C19H20ClNO4/c1-19(2,18(23)24)25-16-9-3-13(4-10-16)11-12-21-17(22)14-5-7-15(20)8-6-14/h3-10H,11-12H2,1-2H3,(H,21,22)(H,23,24)

InChI key

IIBYAHWJQTYFKB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bezafibrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Bezafibrate is a peroxisome proliferator-activated receptor agonist for PPARα, PPARδ, and PPARγ. Lipoprotein lipase (LPL) activator.
The peroxisome proliferator-activated receptor (PPAR) is a member of the steroid nuclear receptor superfamily. Bezafibrate is a peroxisome proliferator-activated receptor agonist for PPARα, PPARδ, and PPARγ. Lipoprotein lipase (LPL) activator.
PPARgamma agonists, including Bezafibrate, have beneficial effects in the suppression of the inflammatory response during RSV infection and therefore might have clinical efficacy in the course of severe RSV-infection.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bezafibrate derivatives as potent effectors of hemoglobin.
C Poyart et al.
Methods in enzymology, 232, 496-513 (1994-01-01)
K L Goa et al.
Drugs, 52(5), 725-753 (1996-11-01)
The lipid-modifying profile of bezafibrate is characterised by marked decreases in elevated triglyceride levels, increases in high density lipoprotein (HDL) cholesterol levels and decreases in total and low density lipoprotein (LDL) cholesterol levels. Bezafibrate also reduces elevated levels of lipoprotein(a)
D Ståhlberg et al.
European journal of clinical pharmacology, 40 Suppl 1, S33-S36 (1991-01-01)
The influence of bezafibrate treatment on hepatic cholesterol metabolism was studied in rats and in humans. The activities of the three key enzymes involved in cholesterol metabolism [3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase, cholesterol 7 alpha-hydroxylase, and acyl-coenzyme A: cholesterol acyltransferase
E Kanterewicz et al.
Annals of the rheumatic diseases, 51(4), 536-538 (1992-04-01)
The case is presented of a 70 year old woman with mild hypercholesterolaemia and hypertension who was readmitted to hospital six months after a previous admission for angina pectoris. The patient was treated with verapamil, nifedipine, and aspirin, and had
U de Faire et al.
Cardiovascular drugs and therapy, 11 Suppl 1, 257-263 (1997-05-01)
A large number of both primary and secondary preventive trials suggest that treatment of elevated plasma lipids may reduce the frequency of coronary heart disease (CHD) events. Meta-analyses indicate that for every 10% reduction of cholesterol, CHD mortality is lowered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service