Skip to Content
Merck
All Photos(1)

Key Documents

A9788

Supelco

(+)-Arabinogalactan

for use with Total Dietary Fiber Control Kit, TDF-C10, from larch wood

Synonym(s):

Polyarabinogalactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

biological source

larch wood

Quality Level

form

lyophilized powder

quality

for use with Total Dietary Fiber Control Kit, TDF-C10

technique(s)

photometry: suitable

mp

>200 °C (dec.) (lit.)

application(s)

food and beverages
general analytical

storage temp.

2-8°C

SMILES string

O1C(C(C(C(C1COC3OCC(C(C3O)OC)O)O)OC)O)OC2C(C(OC(C2O)C)CO)O

InChI

1S/C20H36O14/c1-7-11(23)18(12(24)9(4-21)32-7)34-20-15(27)17(29-3)13(25)10(33-20)6-31-19-14(26)16(28-2)8(22)5-30-19/h7-27H,4-6H2,1-3H3

InChI key

SATHPVQTSSUFFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dietary fiber that enhances immune system function by stimulating natural killer (NK) cell activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaoge Gao et al.
Glycoconjugate journal, 29(4), 159-165 (2012-05-09)
The pH-modified citrus pectin (MCP) has been demonstrated to inhibit galectin-3 in cancer progression. The components and structures of MCP related to this inhibition remained unknown. In this paper, we fractionated MCP on DEAE-cellulose column into a homogenous neutral fraction
N M Mestechkina et al.
Prikladnaia biokhimiia i mikrobiologiia, 47(6), 699-706 (2012-02-01)
The influence of neutral and ionic polysaccharides on the antioxidant (AOA) and detoxifying activities of lactoferrin (LF) and the duration of its circulation in the body was studied. In addition to natural polymers, we studied artificial chitosan derivatives with different
Izabela Pawlaczyk et al.
Carbohydrate polymers, 92(1), 741-750 (2012-12-12)
From the air-dried Wild strawberry (Fragaria vesca L., family Rosaceae) leaves five water-soluble glycoconjugates Fv I-V by different extraction conditions have been isolated. Effects of extraction steps/agents on chemical composition and anticoagulant activity of Fv I-V were examined. Dark brown
Robert W Wheatley et al.
The Journal of biological chemistry, 287(33), 28132-28143 (2012-06-19)
Biosynthesis of the mycobacterial cell wall relies on the activities of many enzymes, including several glycosyltransferases (GTs). The polymerizing galactofuranosyltransferase GlfT2 (Rv3808c) synthesizes the bulk of the galactan portion of the mycolyl-arabinogalactan complex, which is the largest component of the
Ekaterina R Gasilova et al.
The journal of physical chemistry. B, 117(7), 2134-2141 (2013-01-31)
Palladium nanoparticles (PdNPs) are used in catalysis, hydrogen storage, biomedicine, and so on. Arranging the self-assembly of PdNPs within colloidal aggregates is desirable for improving their consumer properties. Stable widely dispersed colloidal aggregates of larch arabinogalactan (LARB) containing nanosized (5-nm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service