Skip to Content
Merck
All Photos(3)

Key Documents

A3139

Sigma-Aldrich

Albumin from mouse serum

lyophilized powder, ≥96% (agarose gel electrophoresis)

Synonym(s):

Mouse albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

mouse

Quality Level

Assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

ELISA: suitable
electrophoresis: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

mouse ... ALB(11657)

Looking for similar products? Visit Product Comparison Guide

General description

Mouse albumin gene is mapped to mouse chromosome 5. It codes for an albumin, which is the most prominent protein in the blood plasma. Albumin is produced by the mature liver. Mouse albumin gene promoter region includes minimum six binding sites (A to F) for specific DNA-binding proteins.

Application

Albumin from mouse serum has been used in labelling by probe 2 for the characterization of matrix metalloprotease-12 by one-dimensional electrophoresis. It has also been used for the preparation of calibrator and controls in enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Albumin facilitates the formation of cytoplasmic lipid droplets in pancreatic stellate cells and adipocyte differentiation of 3T3-L1 cells. In addition, it effectively regulates biological activity of adipogenic transcription factors, peroxisome proliferator-activated receptor (PPAR - γ ) and (CCAAT/enhancer binding protein (C/EBP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Softened food reduces weight loss in the streptozotocin-induced male mouse model of diabetic nephropathy
Norgaard SA, et al.
Laboratory Animals, 52(4), 373-383 (2018)
Albumin-binding domain from Streptococcus zooepidemicus protein Zag as a novel strategy to improve the half-life of therapeutic proteins
Cantante C, et al.
Journal of Biotechnology, 253(6), 23-33 (2017)
Detection of endogenous matrix metalloprotease-12 active form with a novel broad spectrum activity-based probe
Nury C, et al.
The Journal of biological chemistry, 288(8), 5636-5644 (2013)
The interplay of DNA-binding proteins on the promoter of the mouse albumin gene
Lichtsteiner S, et al.
Cell, 51(6), 963-973 (1987)
Marina Pekmezovic et al.
PLoS pathogens, 17(10), e1010037-e1010037 (2021-10-29)
The opportunistic pathogen Candida glabrata is the second most frequent causative agent of vulvovaginal candidiasis (VVC), a disease that affects 70-75% of women at least once during their life. However, C. glabrata is almost avirulent in mice and normally incapable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service