All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
mouse
origin:
USA origin
sterility:
non-sterile
form:
lyophilized powder
Recommended Products
biological source
mouse
sterility
non-sterile
form
lyophilized powder
origin
USA origin
storage temp.
2-8°C
Application
Mouse serum is used in a variety of mouse cell culture systems to study viral infection, inhibition and transduction processes.
Sera from mouse has been used to block non specific antigen binding in immunohistochemistry. It has also been used as a control in AdV neutralizing antibody assay.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Prevalence of neutralizing antibodies against AAV8, AAV9, and AAV843 in a Chinese population
Xu J, et al.
International Journal of Clinical and Experimental Medicine, 12(8), 10253-+-10253-+ (2019)
Brucella ceti associated pathology in the testicle of a harbour porpoise (Phocoena phocoena)
Dagleish MP, et al.
Journal of Comparative Pathology, 139(1), 54-59 (2008)
Suryanarayan Somanathan et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 28(3), 784-793 (2020-02-09)
Intra-arterial administration of an adenovirus serotype 5 (Ad5) vector in a gene therapy trial caused lethal, systemic inflammation in subject 019 with ornithine transcarbamylase deficiency. This unanticipated inflammatory response was absent in another subject receiving the same vector dose and
Elizabeth A Harrington et al.
Human gene therapy, 27(5), 345-353 (2016-01-23)
Isolated methylmalonic acidemia (MMA), a group of autosomal recessive inborn errors of metabolism, is most commonly caused by complete (mut(0)) or partial (mut(-)) deficiency of the enzyme methylmalonyl-CoA mutase (MUT). The severe metabolic instability and increased mortality experienced by many
Anton P McCaffrey et al.
Nature biotechnology, 21(6), 639-644 (2003-05-13)
Hepatitis B virus (HBV) infection substantially increases the risk of chronic liver disease and hepatocellular carcinoma in humans. RNA interference (RNAi) of virus-specific genes has emerged as a potential antiviral mechanism. Here we show that RNAi can be applied to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service