94185
Ethyl maltol
analytical standard
Synonym(s):
2-Ethyl-3-hydroxy-4H-pyran-4-one, Ethyl maltol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
85-95 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCC1=C(O)C(=O)C=CO1
InChI
1S/C7H8O3/c1-2-6-7(9)5(8)3-4-10-6/h3-4,9H,2H2,1H3
InChI key
YIKYNHJUKRTCJL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ethyl maltol is a synthetic homologue of maltol, often found as flavor enhancers and it contributes to the fragrance of commercials products such as cereals, breads, malt beverages, coffee, soybeans and chocolate milk.
Application
Ethyl maltol may be used as an analytical reference standard for the quantification of the analyte in food using spectrophotometric technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Simultaneous kinetic-spectrophotometric determination of maltol and ethyl maltol in food samples by using chemometrics
Food Chemistry, 109(2), 431-438 (2008)
Simultaneous spectrophotometric determination of maltol, ethyl maltol, vanillin and ethyl vanillin in foods by multivariate calibration and artificial neural networks
Food Chemistry, 89(3), 465-473 (2005)
Biochemical pharmacology, 39(6), 1005-1012 (1990-03-15)
The ability of a number of heterocyclic metal chelators to deliver zinc into red cells, to release the liganded zinc to haemoglobin and thereby cause a left shift in the oxygen dissociation curve of intact red cells has been investigated.
Biochemical pharmacology, 37(10), 2051-2057 (1988-05-15)
Accumulation of radioactive iron (59Fe) into isolated fragments of rat small intestine in the presence of two hydroxypyrones, maltol and ethyl maltol, was compared with that in the presence of another chelator of iron(III), nitrilotriacetic acid (NTA). The characteristics of
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 96, 316-323 (2012-06-19)
This paper was designed to investigate the interaction of ethyl maltol with human serum albumin (HSA) under physiological condition by fluorescence, synchronous fluorescence, three-dimensional fluorescence, Fourier transformation infrared spectra, and molecular docking method. Spectroscopic analysis of the emission quenching at
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service