Skip to Content
Merck
All Photos(1)

Key Documents

SCP0054

Sigma-Aldrich

Endothelin Big 1-38 human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C189H282N48O56S5
Molecular Weight:
4282.87
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-His-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Arg-Ser

Application

Endothelin Big (1-38) is the precursor of Endothelin 1. [D-Val22]-Endothelin-1 Big fragment (16-38) is used to inhibit endothelin-converting enzyme activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Hanson et al.
Regulatory peptides, 68(1), 63-69 (1997-01-15)
The hydrolysis of human big endothelin 1-38 (bigET-1) was investigated in the membrane fractions from three human lung specimens. The hydrolysis products were identified by HPLC or by amino acid analysis, peptide sequencing and mass spectrometry, and the contractile effects
Michael Zimmermann et al.
Neurosurgical review, 26(2), 125-132 (2003-09-10)
The aim of this study was to investigate whether the blocking of endothelin-converting enzyme (ECE) activity offers a new approach to inhibiting the development of cerebral vasospasm after subarachnoid hemorrhage (SAH) by preventing transformation of big endothelin-1 (big ET-1) to
A Morita et al.
FEBS letters, 353(1), 84-88 (1994-10-10)
Endothelin converting enzyme (ECE) is essential for generation of the biological effects of endothelin-1 (ET-1) from a precursor, big endothelin-1 (Big ET-1). We synthesized four analogs of human Big ET-1[16-38], substituted with single D-amino acids at P1, P2, P1' and
Marie-Hélène Fecteau et al.
Hypertension (Dallas, Tex. : 1979), 46(1), 87-92 (2005-06-16)
The precursor of endothelin-1, big endothelin-1, can be hydrolyzed by chymase to generate endothelin-1 (1-31) in vitro. In the present study, we explored the processes involved in the production of endothelin-1 (1-31) as well as its pharmacodynamic characteristics in the
Vasileios Kalles et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 33(4), 1231-1236 (2012-03-15)
Deregulation of the endothelin system, comprised of endothelin-1 (ET-1), its isoforms (ET-2 and ET-3) and their receptors (ET(A)R and ET(B)R), is under investigation in various types of human cancer. ET-1 has been suggested to participate in breast cancer development and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service