938998
Sodium hydroxide ChemBeads
Synonym(s):
Caustic soda ChemBeads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
NaHO
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352100
Recommended Products
description
Reagent Type: Inorganic salt
Quality Level
form
solid
composition
loading of base, 14-16 wt. %
reaction suitability
core: sodium
SMILES string
[OH-].[Na+]
InChI
1S/Na.H2O/h;1H2/q+1;/p-1
InChI key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
General description
Sodium hydroxide, also known as lye and caustic soda, is a strong base used universally as a neutralization agent in the chemical industry, paper making, petroleum refining, cleaning compounds, and drain cleaners.
Application
Sodium hydroxide can be used as:
A reagent in the gold-catalyzed selective oxidation of glycerol to glyceric acid. A catalyst for the cross-dehydrogenative coupling of alcohols and hydrosilanes to synthesize silyl ethers.
A base in the Sonogashira-type coupling of aryl halides and alkynes to synthesize ynones.
For general uses, product is also available in powdered form(655104)
A reagent in the gold-catalyzed selective oxidation of glycerol to glyceric acid. A catalyst for the cross-dehydrogenative coupling of alcohols and hydrosilanes to synthesize silyl ethers.
A base in the Sonogashira-type coupling of aryl halides and alkynes to synthesize ynones.
For general uses, product is also available in powdered form(655104)
Features and Benefits
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Other Notes
High-Throughput Reaction Screening with Nanomoles of Solid Reagents Coated on Glass Beads
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Barbara Simonato et al.
Food chemistry, 174, 219-225 (2014-12-23)
The aim of this study was to assess the digestibility of the protein and starch in pasta made with different cereals, i.e. Triticum durum, Triticum polonicum and Triticum dicoccum, and to measure the glycemic index (GI) of the different types
Mathieu André et al.
Organic & biomolecular chemistry, 13(2), 388-397 (2014-11-05)
In the development of our melanoma-selective delivery approach, three preselected conjugates of 5-iodo-2'-deoxyuridine (IUdR) to the ICF01012 melanoma-carrier were radiolabelled with iodine-125, and their in vivo distribution profile was determined. A radioiodination method for the conjugate 1a and its PEGylated
Qianli Zou et al.
Journal of medicinal chemistry, 58(20), 7949-7958 (2015-09-24)
A series of bis(arylidene)cycloalkanone photosensitizers modified by polyethylene glycol (PEG) have been studied for two-photon excited photodynamic therapy (2PE-PDT). As compared with their prototype compounds, these PEGylated photosensitizers show enhanced water solubilities while their photophysical and photochemical properties, including linear
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service