Skip to Content
Merck
All Photos(1)

Key Documents

938971

Sigma-Aldrich

Postassium hydroxide ChemBeads

Synonym(s):

Caustic potash ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KOH
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352100

description

Reagent Type: Inorganic Salt

Quality Level

form

solid

composition

, 14-16 wt. % (loading of base)

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

General description

Potassium hydroxide (KOH) is also known as caustic potash. It is a strong alkali or a solid base compound that can be used in a variety of industrial applications. It finds its usage in the production of soaps, cleaners and detergents. It is frequently used to neutralize acids and prepare potassium salts of reagents.

Application

For general uses, product is also available in powdered form (757551)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eryptosis Indices as a Novel Predictive Parameter for Biocompatibility of Fe3O4 Magnetic Nanoparticles on Erythrocytes
Ran Q, et al.
Scientific reports, 5, 16209-16209 (2015)
Ru-assisted synthesis of Pd/Ru nanodendrites with high activity for ethanol electrooxidation
Zhang K,et.al
Nanoscale, 7(29), 12445-12451 (2015)
Thermochemically activated carbon as an electrode material for supercapacitors
Ostafiychuk B K ,et al.
Nanoscale Research Letters, 10, 65-65 (2015)
Ionic liquid versus prodrug strategy to address formulation challenges
Balk A, et al.
Pharmaceutical Research, 32(6), 2154-2167 (2015)
Franklin Feng Tao et al.
Nature communications, 6, 7798-7798 (2015-08-05)
It is crucial to develop a catalyst made of earth-abundant elements highly active for a complete oxidation of methane at a relatively low temperature. NiCo2O4 consisting of earth-abundant elements which can completely oxidize methane in the temperature range of 350-550 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service