Skip to Content
Merck
All Photos(1)

Key Documents

93302

Supelco

Tris-EDTA buffer solution

BioUltra, for molecular biology, pH 7.4

Synonym(s):

TE buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
ND.02

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.4±0.2 (25 °C)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.01

SMILES string

NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2

InChI key

VLEIUWBSEKKKFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris-EDTA buffer solution has been used for the dilution of DNA samples. It has been used for the elution of purified vector.

Physical form

10mM Trizma® base; 1mM EDTA

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unique nucleotide sequence-guided assembly of repetitive DNA parts for synthetic biology applications.
Torella JP et al.
Nature Protocols, 9, 2075-2075 (2014)
Sin Hang Lee et al.
International journal of environmental research and public health, 16(10) (2019-05-30)
Lyme disease, initially described as Lyme arthritis, was reported before nucleic-acid based detection technologies were available. The most widely used diagnostic tests for Lyme disease are based on the serologic detection of antibodies produced against antigens derived from a single
The effect of COMT Val158Met and DRD2 C957T polymorphisms on executive function and the impact of early life stress.
Klaus K et al.
Brain and Behavior, 7, e00695-e00695 (2017)
Kyla R Rodgers et al.
Scientific reports, 10(1), 7047-7047 (2020-04-29)
Acute inflammation is a key feature of innate immunity that initiates clearance and repair in infected or damaged tissues. Alternatively, chronic inflammation is implicated in numerous disease processes. The contribution of neuroinflammation to the pathogenesis of neurological conditions, including infection
Ying Chen et al.
Applied and environmental microbiology, 74(19), 6132-6137 (2008-08-19)
The relative expression levels of six botulinum neurotoxin cluster genes in a group II Clostridium botulinum type E strain grown at 10 or 30 degrees C were investigated using quantitative real-time reverse transcription-PCR. An enzyme-linked immunosorbent assay was used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service