Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tris-EDTA buffer solution
Synonym(s):
TE buffer solution
Products
Product Category
buffers (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T9285
Tris-EDTA buffer solution,
100X concentrate for Northern and Southern blotting
Expand
View Pricing
93283
Tris-EDTA buffer solution,
BioUltra, for molecular biology, pH 8.0
Expand
View Pricing
93302
Tris-EDTA buffer solution,
BioUltra, for molecular biology, pH 7.4
Expand
View Pricing