Skip to Content
Merck
All Photos(1)

Key Documents

918911

Sigma-Aldrich

4-Methyl-4-mercaptopentan-2-one

≥95.0%

Synonym(s):

4-Mercapto-4-methyl-2-pentanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12OS
CAS Number:
Molecular Weight:
132.22
MDL number:
UNSPSC Code:
12352105
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

≥95.0%

form

liquid

functional group

ketone
thiol

storage temp.

2-8°C

SMILES string

CC(=O)CC(C)(C)S

InChI

1S/C6H12OS/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3

InChI key

QRNZMFDCKKEPSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z744036PL800ASSPIEUBMSW3120
feature

CE compliant

feature

-

feature

CE compliant

feature

-

manufacturer/tradename

Plas-Labs 800-AS/SPI/CE/UK

manufacturer/tradename

Plas-Labs 800AS/SPI/J

manufacturer/tradename

Plas-Labs 800-AS/SPI/CE/EU

manufacturer/tradename

Benchmark Scientific W3120

Overall H × W × depth

5.0 in. × 4 1/4 in. × 2 1/2 in.

Overall H × W × depth

5.0 in. × 4 1/4 in. × 2 1/2 in.

Overall H × W × depth

5.0 in. × 4 1/4 in. × 2 1/2 in.

Overall H × W × depth

-

weight

2 lb

weight

2 lb

weight

2 lb

weight

-

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service