Skip to Content
Merck
All Photos(1)

Key Documents

88652

Supelco

Thioglycolic acid solution

~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites

Synonym(s):

Mercaptoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOH
CAS Number:
Molecular Weight:
92.12
Beilstein:
506166
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

quality

for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites

concentration

~80% in H2O

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.471

density

1.265 g/mL at 20 °C

SMILES string

OC(=O)CS

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thioglycolic acid is an organic colourless compound with thiol and carboxylic acid as functional group. It is mainly used to make depilatories. It is considered to be corrosive to metals and tissues.

Application

Mercaptoacetic acid has been used as a stabilizer for preparation of silver colloids, in a UV-vis spectroscopy study to establish stable and highly surface-enhanced Raman spectroscopy (SERS)-active silver colloids.

Other Notes

Reagent for the extraction and spectrophotometric determination of various transition metals, e.g. Pd; W; Mo; Ti

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercaptoacetic acid-capped silver nanoparticles colloid: formation, morphology, and SERS activity.
Li, Xiaoling, et al.
Langmuir, 19.10, 4285-4290 (2003)
S Banerjee
Talanta, 33(4), 360-362 (1986-04-01)
A sensitive spectrophotometric method for the determination of titanium by formation of its complex with tannin and thioglycollic acid at pH 4 has been developed. The intense yellow colour is measured at 400 nm and the system obeys Beer's law
S.P. Rao et al.
Indian J. Chem. A, 20, 639-639 (1981)
G. Wunsch et al.
Fresenius Journal of Analytical Chemistry, 295, 119-119 (1979)
A. Diamantatos
Analytica Chimica Acta, 61, 233-233 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service