Skip to Content
Merck
All Photos(1)

Key Documents

87139

Supelco

Tetradecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH3
CAS Number:
Molecular Weight:
198.39
Beilstein:
1733859
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6.83 (vs air)

vapor pressure

1 mmHg ( 76.4 °C)

description

olefine free

Assay

≥99% (GC)

autoignition temp.

455 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

252-254 °C (lit.)

mp

5.5 °C (lit.)

density

0.762 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetradecane is a straight-chain alkane, classified as a volatile organic compound (VOC).

Application

The analytical standard can also be used as follows:

  • Determination of seven volatile organic compounds in the surgically resected colonic tissue samples of colorectal cancer patients by gas chromatography-mass spectrometry (GC-MS) following their extraction by headspace- and direct injection- based solid phase microextraction (SPME)
  • Measurement of concentrations of volatile organic compounds (VOCs) in biogas samples obtained from three different biogas plants by gas chromatography-mass spectrometry
  • Simultaneous identification and determination of volatile organic compounds (VOCs) and semi-volatile organic compounds (SVOCs) from air samples using a styrene-divinylbenzene polymer particle and activated carbon particles based sampling device and GC-MS
  • Development and validation of a thermal desorption method coupled with gas chromatography-mass spectrometry (GC-MS) for the determination of VOCs in biogas samples after their collection into inert film bags for sample treatment in multi-sorbent bed tubes
  • Gas chromatography-mass spectrometry (GC-MS) method-based profiling of volatile organic compounds in the olive fruit headspace extracts

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Transcendental Meditation Program for Business People (1976)
Agnieszka Ewa Wiącek
Colloids and surfaces. B, Biointerfaces, 83(1), 108-115 (2010-12-07)
The properties of n-tetradecane/electrolyte emulsions with DPPC or DPPC vesicles in the electrolyte solution were investigated. The DPPC molecules form different aggregates, which possess different surface affinity, size and structure, and therefore we assumed some differences in the adsorption at
Christoph Aeppli et al.
Environmental science & technology, 43(23), 8813-8820 (2009-12-01)
Mass transfer of organic contaminants from nonaqueous phase liquids to the aqueous phase can significantly modulate the observable carbon isotope fractionation behavior associated with contaminant transformation. We evaluated the effects of kinetic interphase mass transfer between tetradecane and water on
Stephan Kubowicz et al.
Langmuir : the ACS journal of surfaces and colloids, 25(2), 952-958 (2009-01-30)
The behavior of mixed-ligand-coated gold nanoparticles at a liquid-liquid interface during compression has been investigated. The system was characterized by measuring pressure-area isotherms and by simultaneously performing in situ X-ray studies. Additionally, Monte Carlo (MC) simulations were carried out in
Robert Byrne et al.
Physical chemistry chemical physics : PCCP, 12(8), 1895-1904 (2010-02-11)
Investigations into the extent of structuring present in phosphonium based ionic liquids (ILs) have been carried out using photochromic molecular probes. Three spiropyran derivatives containing hydroxyl (BSP-1), carboxylic acid (BSP-2) and aliphatic chain (C(14)H(29)) (BSP-3) functional groups have been analysed

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service