Skip to Content
Merck
All Photos(1)

Key Documents

86478

Sigma-Aldrich

γ-Terpinene

≥95.0% (GC)

Synonym(s):

gamma-Terpinene, 1-Isopropyl-4-methyl-1,4-cyclohexadiene, p-Mentha-1,4-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2038347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

Quality Level

vapor pressure

~0.7 mmHg ( 20 °C)

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.474 (lit.)
n20/D 1.474

bp

182 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

SMILES string

CC(C)C1=CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,7-8H,5-6H2,1-3H3

InChI key

YKFLAYDHMOASIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Cassia, lime

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

124.9 °F - closed cup

Flash Point(C)

51.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad S Abu-Darwish et al.
Natural product research, 26(14), 1310-1317 (2011-06-29)
The effect of plant space and time of harvesting on yield and quality of Thymus vulgaris was evaluated in Jordan. Thyme was cultivated in rows of 50 cm apart with inter-row spacing of 15, 30 or 45 cm and was grown at
D Djenane et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 17(6), 505-515 (2011-11-04)
Essential oils (EOs) extracted by hydrodistillation from leaf parts of Algerian Eucalyptus globulus, Myrtus communis and Satureja hortensis were analyzed by gas chromatography/mass spectrometry (GC/MS). The main components of EOs obtained were γ-terpinene (94.48%), 1,8-cineole (46.98%) and carvacrol (46.10%), respectively
Ekhteraei Tousi Samaneh et al.
Natural product research, 24(19), 1834-1842 (2010-11-26)
Essential oils from the subterranean organs of three species of Valeriana L. from Iran (Valeriana sisymbriifolia Vahl, Valeriana alliariifolia Adams and Valeriana officinalis L.) belonging to Valerianaceae family have been obtained by hydrodistillation and analysed by gas chromatography-mass spectrometry in
Wei-bin Yuan et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1067-1069 (2011-11-10)
To analysis the constituents of volatile oil from Fructus Auranti Immaturus by GC-MS. The volatile oil was extracted by steam distillation, then separated by capillary gas chromatography. The constituents of volatile oil were identified and their amount were determined by
Xiao-Dong Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(10), 1544-1548 (2012-03-01)
To extract the essential oils from the Seedlings, the Aseptic Seedlings and the Tissue Culture Seedlings of Thymus vulgaris and analyze their chemical components and the relative contents. The essential oils were extracted by steam distillation, the chemical components and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service