Merck
  • Chemical composition and antimicrobial effects of essential oils of Eucalyptus globulus, Myrtus communis and Satureja hortensis against Escherichia coli O157:H7 and Staphylococcus aureus in minced beef.
Chemical composition and antimicrobial effects of essential oils of Eucalyptus globulus, Myrtus communis and Satureja hortensis against Escherichia coli O157:H7 and Staphylococcus aureus in minced beef.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional (2011-11-04)
D Djenane, J Yangüela, T Amrouche, S Boubrit, N Boussad, P Roncalés
ABSTRACT
Essential oils (EOs) extracted by hydrodistillation from leaf parts of Algerian Eucalyptus globulus, Myrtus communis and Satureja hortensis were analyzed by gas chromatography/mass spectrometry (GC/MS). The main components of EOs obtained were γ-terpinene (94.48%), 1,8-cineole (46.98%) and carvacrol (46.10%), respectively, for E. globulus, M. communis and S. hortensis. The in vitro antimicrobial activity of the EOs was evaluated against Staphylococcus aureus CECT 4459 and Escherichia coli O157:H7 CECT 4267 using the agar diffusion technique. Results revealed that E. globulus and S. hortensis EOs had more antibacterial effects than that from M. communis. Minimal inhibitory concentrations (MIC) showed a range of 0.05-0.22% (volume by volume [v/v]). Sensitivity of gram-positive S. aureus was much higher than that of gram-negative E. coli. Plant EOs were added to minced beef (two-fold MIC value) at 0.10-0.44%, experimentally inoculated with the same pathogens at a level of 5 × 10(5) colony forming units (cfu)/g and stored at 5 ± 2 °C. Results showed that the EOs of E. globulus and S. hortensis had remarkable antibacterial properties, higher than that of M. communis, against S. aureus and E. coli. Indeed, a reduction of 5.8 log cfu/g (70.74% of reduction) was recorded after 7 days of storage for S. hortensis against E. coli. However, regarding S. aureus, both S. hortensis and E. globulus caused a highly significant (p < 0.05) decrease of microbial counts, most evident after 5 days of storage; S. aureus numbers were 3.50 and 2.50 cfu/g, respectively, corresponding to a reduction of 2.20 and 3.20 log cfu/g (38.60 and 56.14% of reduction) after 1 week of storage. Sensory evaluation revealed that the aroma of minced beef meat treated with EOs was acceptable by panelists at the levels used.
Sigma-Aldrich
γ-Terpinene, 97%
Sigma-Aldrich
α-Terpinene, 85%
Sigma-Aldrich
γ-Terpinene, ≥95%, FG
Sigma-Aldrich
α-Terpinene, ≥89%, FCC, FG
Sigma-Aldrich
α-Terpinene, ≥95.0% (GC)
Supelco
γ-Terpinene, analytical standard
Sigma-Aldrich
γ-Terpinene, ≥95.0% (GC)
Sigma-Aldrich
γ-Terpinene, natural, 95%, FG