Salicylic acid
Synonym(s):
Salicylic acid, 2-Hydroxybenzoic acid
Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing