Skip to Content
Merck
All Photos(1)

Key Documents

81293

Supelco

Poly(ethylene glycol)

analytical standard, for GPC, 6,000

Synonym(s):

PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~5850
Mp ~6240
Mw ~6000

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.03

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Quantifying thiolated chemical additives for copper electroplating process: Demonstrates the use of Poly(ethylene glycol) (PEG) in enhancing the performance of copper electroplating, a critical application in electronic manufacturing (Wang et al., 2024).

  • Novel fibrous Ag(NP) decorated clay-polymer composite: Highlights PEG′s role in developing composites for water purification, addressing the urgent need for efficient removal of micro-pollutants and bacteria from water sources (Goswami et al., 2024).

  • Phenylacetylene-Terminated Poly(Ethylene Glycol) as Ligands for Colloidal Noble Metal Nanoparticles: Introduces a novel PEG-based ligand system for noble metal nanoparticles, advancing applications in catalysis and materials science (Duan et al., 2024).

  • Assessing the Effect of Surface Coating on the Stability, Degradation, Toxicity and Cell Endocytosis/Exocytosis of Upconverting Nanoparticles: Explores the role of PEG in modifying nanoparticles to enhance their biocompatibility and stability, crucial for drug delivery and diagnostic applications (Arellano et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
P I Polimeni et al.
Journal of cardiovascular pharmacology, 14(3), 374-380 (1989-09-01)
The acute hemodynamic effects of an intravenously (i.v.) injected poly(ethylene oxide), Polyox WSR N-60K (dose 50 mg/kg), were studied in the open-chest rat anesthetized with sodium pentobarbital. The injectate is one of four drag-reducing polymers known to augment in vitro
Mark A Rice et al.
Acta biomaterialia, 5(1), 152-161 (2008-09-17)
Ultrasound has potential as a non-destructive analytical technique to provide real-time online assessments of matrix evolution in cell-hydrogel constructs used in tissue engineering. In these studies, chondrocytes were encapsulated in poly(ethylene glycol) hydrogels, and gel degradation was manipulated to provide
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service