Skip to Content
Merck
All Photos(2)

Key Documents

81153

Supelco

Poly(dimethylsiloxane)

analytical standard, for GPC, 311

Synonym(s):

Dimethylpolysiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Si(CH3)2O]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

vapor pressure

5 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~311
Mp ~311
Mw ~311

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.0

format

neat

storage temp.

2-8°C

SMILES string

C[Si](C)=O

Looking for similar products? Visit Product Comparison Guide

General description

Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.

Application

It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
Poly(dimethylsiloxane) is found to be highly compatible with organic solvents and is used for fabricating microfluidic devices..

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent compatibility of poly (dimethylsiloxane)-based microfluidic devices.
Lee NGJ, et al.
Analytical Chemistry, 75(23), 6544-6554 (2003)
Robust and biodegradable elastomers based on corn starch and polydimethylsiloxane (PDMS).
Ceseracciu L
ACS Applied Materials & Interfaces, 7(6), 3742-3753 (2015)
Molecular weight distribution of poly (dimethylsiloxane) by combining matrix-assisted laser desorption/ionization time-of-flight mass spectrometry with gel-permeation chromatography fractionation.
Montaudo, Giorgio, et al.
Rapid Communications in Mass Spectrometry, 9.12, 1158-1163 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service