Skip to Content
Merck
All Photos(1)

Key Documents

81091

Sigma-Aldrich

Dextromethorphan hydrobromide monohydrate

tested according to Ph. Eur.

Synonym(s):

Dextromethorphan hydrobromicum, (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32
Beilstein:
6453793
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

Br[H].[H]O[H].[H][C@]12CCCC[C@]13CCN(C)[C@H]2Cc4ccc(OC)cc34

InChI

1S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H;1H2/t15-,17+,18+;;/m1../s1

InChI key

STTADZBLEUMJRG-IKNOHUQMSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fixed drug eruption due to dextromethorphan with tolerance to other opioids.
G Davila-Fernández et al.
Journal of investigational allergology & clinical immunology, 23(4), 281-282 (2013-08-24)
Tao-Cheng Wu et al.
PloS one, 7(9), e46067-e46067 (2012-10-11)
Vascular oxidative stress may be increased with age and aggravate endothelial dysfunction and vascular injury in hypertension. This study aimed to investigate the effects of dextromethorphan (DM), a NADPH oxidase inhibitor, either alone or in combination treatment, on blood pressure
Melanie A Paquette et al.
The European journal of neuroscience, 36(9), 3224-3234 (2012-08-07)
Amantadine and dextromethorphan suppress levodopa (L-DOPA)-induced dyskinesia (LID) in patients with Parkinson's disease (PD) and abnormal involuntary movements (AIMs) in the unilateral 6-hydroxydopamine (6-OHDA) rat model. These effects have been attributed to N-methyl-d-aspartate (NMDA) antagonism. However, amantadine and dextromethorphan are
F L Opdam et al.
Cancer chemotherapy and pharmacology, 71(3), 593-601 (2012-12-12)
Adjuvant therapy with tamoxifen significantly reduces breast cancer recurrence and mortality in estrogen receptor positive disease. CYP2D6 is the main enzyme involved in the activation of the prodrug tamoxifen into the anti-estrogen endoxifen. Endoxifen is thought to be a main
Ewa Tomczak et al.
Archiwum medycyny sadowej i kryminologii, 62(3), 197-202 (2013-05-09)
The study highlighted the problem of intoxication using substances and/or preparations, to which nowadays young people have unrestricted access. Based on the case developed in the Department of Forensic Medicine of the Medical University of Gdansk, our team members were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service