Skip to Content
Merck
All Photos(1)

Key Documents

80430

Sigma-Aldrich

Dichlorodimethylsilane

produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)

Synonym(s):

Silane M2, DMDCS, Dimethyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SiCl2
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

70 °C (lit.)

mp

−76 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlorodimethylsilane is an organosilicon compound generally used as a protecting group for diols and carbonyl compounds. It is also used as a starting material for the preparation of silicon-based reagents.

Application

Dichlorodimethylsilane can be used:
  • As a silane coupling agent in the synthesis of silica nanoparticles and in their surface modification studies.
  • To prepare polydimethylsiloxane microemulsions by reacting with sodium dodecylpolyoxyethylene sulfate.
  • To prepare a novel heterasumanene where the benzylic carbon atoms of the sumanene are replaced by heteroatom (S, Si) functionalities

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongyan Lu et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(5), R558-R570 (2014-06-13)
Ruminal fermentation products such as short-chain fatty acids (SCFA) and CO2 acutely stimulate urea transport across the ruminal epithelium in vivo, whereas ammonia has inhibitory effects. Uptake and signaling pathways remain obscure. The ruminal expression of SLC14a1 (UT-B) was studied
R Bos et al.
Microbiology (Reading, England), 142 ( Pt 9), 2355-2361 (1996-09-01)
Co-adhesion between oral microbial pairs (i.e. adhesion of a planktonic microorganism to a sessile organism adhering to a substratum surface) has been described as a highly specific interaction, mediated by stereochemical groups on the interacting microbial cell surfaces, and also
Wonmi Ahn et al.
ACS nano, 4(7), 4181-4189 (2010-06-23)
Uniform hexagonal arrays of diverse nanotemplated metal structures were formed via selective electroless gold plating on particle-lithographed dimethyldichlorosilane layers. Surface-associated water at silica bead interstices was shown to correlate with the formation of silane rings with outer ring diameters ranging
Zhan-Hui Wang et al.
Journal of immunological methods, 285(2), 237-243 (2004-02-26)
In the development of biosensors, the immobilization of biomolecules at interfaces played a crucial role. The feasibility of using 3-aminopropyltriethoxysilane (APTES) and glutaraldehyde (Glu) to modify silicon surface to immobilize covalently protein for immunoassay with the biosensor based on imaging
S N Krylov et al.
Electrophoresis, 21(4), 767-773 (2000-03-25)
Capillary electrophoresis (CE) is an important tool of chemical cytometry. Whole-cell analysis using CE starts with cell injection into the capillary by either siphoning or electroosmosis. However, strong adherence of the cell to the support surface can prevent efficient cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service