Skip to Content
Merck
All Photos(1)

Key Documents

D61504

Sigma-Aldrich

Dichlorodiphenylsilane

97%

Synonym(s):

Diphenyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2SiCl2
CAS Number:
Molecular Weight:
253.20
Beilstein:
609882
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Quality Level

vapor pressure

2 mmHg ( 125 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.578 (lit.)

bp

305 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C12H10Cl2Si/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

OSXYHAQZDCICNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlorodiphenylsilane is a silane based surface modifier, and a precursor which can be used in the synthesis of silica based materials. It is used to modify the surface by providing toughness, durability and resistance to shock.

Application

Dichlorodiphenylsilane can be used in the synthesis of a host material for the fabrication of blue phosphorescent organic light emitting diodes (OLEDs).
The addition reaction of dichlorodiphenylsilane with diglycidyl ether of bisphenol-S yields poly(silyl ether) with pendant chloromethyl groups. {40} The coupling reaction between dichloromethylsilane and dichlorodiphenylsilane can form diorganosilane-hydrosilane copolymers at -75oC. Heating the compound with tetrabutoxydianilinocyclophosphazene at temperatures between 100oC-160oC may yield cyclophosphazene oligomers.{42}

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of poly (silyl ether) by the addition reaction of bisphenol-S diglycidyl ether and dichlorodiphenylsilane.
Liaw DJ
Polymer, 38(20), 5217-5219 (1997)
Preparation and Properties of Polysiloxane Modified Epoxy Encapsulating Material
Advanced Materials Research, 1021(6), 3-6 (2014)
Diphenylsilanes containing electronically isolated carbazolyl fragments as host materials for light emitting diodes
Zostautiene R, et al.
Synthetic Metals, 161(1-2), 92-95 (2011)
Si-H functional polysilanes via a homogeneous reductive coupling reaction.
Sacarescu G, et al.
Macromolecular Rapid Communications, 22(6), 405-408 (2001)
Branched diphenylsilane derivatives containing electronically isolated indolyl moieties as host materials for blue organic light emitting diodes
Zostautiene R, et al.
Dyes and Pigments, 91(2), 177-181 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service