72090
Neocuproine hydrochloride monohydrate
for spectrophotometric det. of Cu, ≥99.0%
Synonym(s):
2,9-Dimethyl-1,10-phenanthroline hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H12N2 · HCl · H2O
CAS Number:
Molecular Weight:
262.73
Beilstein:
4584190
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Quality Level
Assay
≥99.0% (AT)
≥99.0%
form
powder
quality
for spectrophotometric det. of Cu
technique(s)
UV/Vis spectroscopy: suitable
solubility
H2O: 25 mg/mL, clear
SMILES string
Cl[H].[H]O[H].Cc1ccc2ccc3ccc(C)nc3c2n1
InChI
1S/C14H12N2.ClH.H2O/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9;;/h3-8H,1-2H3;1H;1H2
InChI key
APDYLFLZNGECIM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Neocuproine hydrochloride monohydrate has been used in measuring copper dissolution in Cu–Ni alloys using colorimetric method. It has also been used in preparing complexing agent solution in studying the antioxidant capacity assays in biological samples based on Cu reducing complexes.
Neocuproine hydrochloride monohydrate has been used in measuring copper dissolution in Cu–Ni alloys using colorimetric method. It has also been used in preparing complexing agent solution in studying the antioxidant capacity assays in biological samples based on Cu reducing complexes. It may be used as a precursor for the preparation of neocuproine, used as an analytical reagent for the determination of copper in environmental samples using spectrophotometric technique.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Corrosion inhibitive action of cysteine on Cu?30Ni alloy in aerated 0.5 MH 2 SO 4.
Saifi, H., et al.
Materials Chemistry and Physics, 120.2, 661-669 (2010)
Rapid speciation of trace iron in rainwater by reverse flow injection analysis coupled to a long path length liquid waveguide capillary cell and spectrophotometric detection
Huang Y, et al.
Microchimica Acta, 166(3-4), 221-228 (2008)
Insights on antioxidant assays for biological samples based on the reduction of copper complexes?the importance of analytical conditions.
Marques, Sara S., et al.
International Journal of Molecular Sciences, 15.7, 11387-11402 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service