Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium selenite
Linear Formula:
Na
2
SeO
3
CAS Number:
10102-18-8
Molecular Weight:
172.94
EC Number:
233-267-9
Products
Product Category
salts (4)
media supplements (1)
Brand
Sigma-Aldrich (4)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Purum (1)
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
S5261
Sodium selenite,
BioReagent, suitable for cell culture, ≥98%
Expand
View Pricing
214485
Sodium selenite,
99%
Expand
View Pricing
S9133
Sodium selenite,
γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Expand
View Pricing
71950
Sodium selenite,
anhydrous, ≥90.0% (RT)
Expand
View Pricing