Skip to Content
Merck
All Photos(2)

Key Documents

65839

Supelco

Ginsenoside Rf

analytical standard

Synonym(s):

(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, Panaxoside Rf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H][C@]1(O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@@]2([H])O[C@H]3C[C@]4(C)[C@]([H])(C[C@@H](O)[C@]5([H])[C@]([H])(CC[C@@]45C)[C@@](C)(O)CC\C=C(/C)C)[C@@]6(C)CC[C@H](O)C(C)(C)[C@]36[H]

InChI

1S/C42H72O14/c1-20(2)10-9-13-42(8,52)21-11-15-40(6)28(21)22(45)16-26-39(5)14-12-27(46)38(3,4)35(39)23(17-41(26,40)7)53-37-34(32(50)30(48)25(19-44)55-37)56-36-33(51)31(49)29(47)24(18-43)54-36/h10,21-37,43-52H,9,11-19H2,1-8H3/t21-,22+,23-,24+,25+,26+,27-,28-,29+,30+,31-,32-,33+,34+,35-,36-,37+,39+,40+,41+,42-/m0/s1

InChI key

UZIOUZHBUYLDHW-XUBRWZAZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohit Mahar et al.
Scientific reports, 10(1), 8885-8885 (2020-06-03)
Increased glucose uptake and aerobic glycolysis are striking features of many cancers. These features have led to many techniques for screening and diagnosis, but many are expensive, less feasible or have harmful side-effects. Here, we report a sensitive 1H/2H NMR
Cuk-seong Kim et al.
Acta pharmacologica Sinica, 26(5), 551-558 (2005-04-22)
To examine the effects of ginseng saponins on the heterologously expressed human ether-a-go-go related gene (HERG) that encodes the rapid component of the delayed rectifier K+ channel. A two-electrode voltage clamp technique was used. HERG currents were recorded in Xenopus
Noel S Quiming et al.
Analytical and bioanalytical chemistry, 389(5), 1477-1488 (2007-09-07)
The influences of the organic component of the mobile phase and the column temperature on the retention of ginsenosides on a poly(vinyl alcohol) (PVA) bonded stationary phase operated under hydrophilic interaction chromatographic mode were investigated. The retention of the ginsenosides
Sunoh Kim et al.
Biochemical and biophysical research communications, 365(3), 399-405 (2007-11-21)
In the present study, we have examined any possible involvement of L-type Ca(2+) channels in ginseng-mediated neuroprotective actions. Exposure to a 50mM KCl (high-K) produced neuronal cell death, which was blocked by a selective L-type Ca(2+) channel blocker in cultured
The antimetastatic and immunomodulating activities of ginseng minor glycosides.
A M Popov et al.
Doklady. Biochemistry and biophysics, 380, 309-312 (2001-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service