Skip to Content
Merck
All Photos(1)

Key Documents

G-015

Supelco

Ginseng Ginsenosides Mix

100 μg/mL each component in methanol, ampule of 100 μg/mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O13
Molecular Weight:
785.01
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Ginsenoside Rb1
Ginsenoside Rb2
Ginsenoside Rc
Ginsenoside Rd
Ginsenoside Re
Ginsenoside Rf
Ginsenoside Rg1
Ginsenoside Rg2

packaging

ampule of 100 μg/mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL each component in methanol

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

A certified multi-component mix of the following Ginseng Ginsenosides in methanol: Rb1, Rb2, Rc, Rd, Re, Rf, Rg1, and Rg2. This Snap-N-Shoot® solution mix is suitable for quality control testing of Ginseng dietary supplements by LC or LC/MS. The Snap-N-Shoot® format and multi-component solution mix offer several advantages from added convenience with less time weighing plant extracts or powders and preparing solutions to improved efficiency from less time rerunning samples or remaking reference solutions due to different raw material purity factors. The sealed ampoule provides additional precautions for storage of these sensitive phytochemicals. genus panax

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service